Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24827763-24828012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY3
Entrez Gene ID
109
Gene Name
adenylate cyclase 3
Gene Aliases
AC-III, AC3
UniGene
Hs.467898
Gene Chromosome Location
Chr.2: 24819169-24920017 on Build GRCh38
Transcripts
NM_001320613.1 NM_004036.4 XM_006711925.1 XM_011532489.1 XM_011532490.1 XM_011532491.1 XM_011532492.2 XM_011532493.1 XM_011532494.1 XM_011532496.1 XM_017003186.1 XM_017003187.1 XM_017003188.1 XM_017003189.1 XM_017003190.1 XM_017003191.1 XM_017003192.1 XM_017003193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4618216 c.2406C>T p.Y802Y Chr.2: 24827928-24827928 on Build GRCh38
1283292 c.2338A>T p.M780L Chr.2: 24827996-24827996 on Build GRCh38
6157745 c.2331C>T p.L777L Chr.2: 24828003-24828003 on Build GRCh38
6594911 c.2391C>T p.P797P Chr.2: 24827943-24827943 on Build GRCh38
2911549 c.2431G>A p.D811N Chr.2: 24827903-24827903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759333413 Chr.2: 24827857-24827857 on Build GRCh38
rs765173644 Chr.2: 24827962-24827962 on Build GRCh38
rs146269706 Chr.2: 24827852-24827852 on Build GRCh38
rs775904787 Chr.2: 24827945-24827945 on Build GRCh38
rs1045815907 Chr.2: 24827828-24827828 on Build GRCh38
rs767110396 Chr.2: 24827885-24827885 on Build GRCh38
rs565303281 Chr.2: 24827889-24827889 on Build GRCh38
rs759846027 Chr.2: 24827880-24827880 on Build GRCh38
rs749363565 Chr.2: 24827927-24827927 on Build GRCh38
rs774738640 Chr.2: 24827937-24827937 on Build GRCh38
rs1045870709 Chr.2: 24827952-24827952 on Build GRCh38
rs758474445 Chr.2: 24827975-24827975 on Build GRCh38
rs140426660 Chr.2: 24827966-24827966 on Build GRCh38
rs201034328 Chr.2: 24827870-24827870 on Build GRCh38
rs753347971 Chr.2: 24827903-24827903 on Build GRCh38
rs779982153 Chr.2: 24827976-24827976 on Build GRCh38
rs532463063 Chr.2: 24827892-24827892 on Build GRCh38
rs779412964 Chr.2: 24827987-24827987 on Build GRCh38
rs772629343 Chr.2: 24827990-24827990 on Build GRCh38
rs763500351 Chr.2: 24827876-24827876 on Build GRCh38
rs752824508 Chr.2: 24827867-24827870 on Build GRCh38
rs777026103 Chr.2: 24827865-24827865 on Build GRCh38
rs377506465 Chr.2: 24827928-24827928 on Build GRCh38
rs372148762 Chr.2: 24827853-24827853 on Build GRCh38
rs373035912 Chr.2: 24827918-24827918 on Build GRCh38
rs897855322 Chr.2: 24827831-24827831 on Build GRCh38
rs764980386 Chr.2: 24827872-24827872 on Build GRCh38
rs542868330 Chr.2: 24827822-24827822 on Build GRCh38
rs767804097 Chr.2: 24827862-24827864 on Build GRCh38
rs770197739 Chr.2: 24827920-24827920 on Build GRCh38
rs543479985 Chr.2: 24827863-24827863 on Build GRCh38
rs778757630 Chr.2: 24827906-24827906 on Build GRCh38
rs773315426 Chr.2: 24827922-24827922 on Build GRCh38
rs369803323 Chr.2: 24827942-24827942 on Build GRCh38
rs537829337 Chr.2: 24827986-24827986 on Build GRCh38
rs758279809 Chr.2: 24827912-24827912 on Build GRCh38
rs1028116101 Chr.2: 24827824-24827824 on Build GRCh38
rs772433588 Chr.2: 24827943-24827943 on Build GRCh38
rs1008087534 Chr.2: 24827971-24827971 on Build GRCh38
rs757211680 Chr.2: 24827856-24827856 on Build GRCh38
rs991120544 Chr.2: 24827941-24827941 on Build GRCh38
rs576369380 Chr.2: 24827854-24827854 on Build GRCh38
rs190125602 Chr.2: 24827948-24827948 on Build GRCh38
rs766839931 Chr.2: 24827877-24827877 on Build GRCh38
rs572315516 Chr.2: 24827796-24827796 on Build GRCh38
rs185180064 Chr.2: 24827917-24827917 on Build GRCh38
rs761261963 Chr.2: 24827958-24827958 on Build GRCh38
rs749337506 Chr.2: 24827984-24827984 on Build GRCh38
rs560339786 Chr.2: 24827911-24827911 on Build GRCh38
rs934311006 Chr.2: 24827922-24827924 on Build GRCh38
rs372560995 Chr.2: 24827947-24827947 on Build GRCh38
rs777887022 Chr.2: 24827983-24827983 on Build GRCh38
rs373053710 Chr.2: 24827869-24827869 on Build GRCh38
rs755558661 Chr.2: 24827890-24827890 on Build GRCh38
rs756369092 Chr.2: 24827982-24827982 on Build GRCh38
rs919336331 Chr.2: 24827963-24827963 on Build GRCh38
rs139420185 Chr.2: 24827904-24827904 on Build GRCh38
rs78678013 Chr.2: 24827978-24827978 on Build GRCh38
rs774744041 Chr.2: 24827878-24827878 on Build GRCh38

View Full Product Details