Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137251549-137251796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf173-AS1
Entrez Gene ID
100129722
Gene Name
C9orf173 antisense RNA 1
Gene Aliases
-
UniGene
Hs.656347
Gene Chromosome Location
Chr.9: 137250219-137253497 on Build GRCh38
Transcripts
NR_038389.1 NM_001004353.3 NM_001256699.1 NM_001256700.1 NM_001256701.1 NR_046339.1 NR_046340.1 XM_006717113.2 XM_006717115.2

Gene Symbol
STPG3
Entrez Gene ID
441476
Gene Name
sperm-tail PG-rich repeat containing 3
Gene Aliases
C9orf173
UniGene
Hs.372640
Gene Chromosome Location
Chr.9: 137250840-137253483 on Build GRCh38
Transcripts
NR_038389.1 NM_001004353.3 NM_001256699.1 NM_001256700.1 NM_001256701.1 NR_046339.1 NR_046340.1 XM_006717113.2 XM_006717115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1461368 c.146C>T p.P49L Chr.9: 137251773-137251773 on Build GRCh38
5612953 c.150G>A p.G50G Chr.9: 137251777-137251777 on Build GRCh38
3716264 c.140C>T p.P47L Chr.9: 137251767-137251767 on Build GRCh38
6290424 c.111-9C>T p.? Chr.9: 137251729-137251729 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs554611019 Chr.9: 137251696-137251696 on Build GRCh38
rs1024571507 Chr.9: 137251630-137251630 on Build GRCh38
rs942240247 Chr.9: 137251683-137251683 on Build GRCh38
rs867548311 Chr.9: 137251740-137251740 on Build GRCh38
rs948663900 Chr.9: 137251764-137251764 on Build GRCh38
rs573010517 Chr.9: 137251572-137251572 on Build GRCh38
rs960546150 Chr.9: 137251632-137251632 on Build GRCh38
rs200942039 Chr.9: 137251751-137251751 on Build GRCh38
rs544039067 Chr.9: 137251671-137251671 on Build GRCh38
rs191532150 Chr.9: 137251646-137251646 on Build GRCh38
rs762442322 Chr.9: 137251711-137251711 on Build GRCh38
rs890798843 Chr.9: 137251743-137251743 on Build GRCh38
rs538917855 Chr.9: 137251586-137251586 on Build GRCh38
rs558629622 Chr.9: 137251645-137251645 on Build GRCh38
rs1037986147 Chr.9: 137251752-137251752 on Build GRCh38
rs778549492 Chr.9: 137251757-137251757 on Build GRCh38
rs957389795 Chr.9: 137251669-137251669 on Build GRCh38
rs1038082600 Chr.9: 137251694-137251694 on Build GRCh38
rs1049358165 Chr.9: 137251727-137251727 on Build GRCh38
rs982531566 Chr.9: 137251750-137251750 on Build GRCh38
rs767716789 Chr.9: 137251716-137251716 on Build GRCh38
rs1008527473 Chr.9: 137251745-137251745 on Build GRCh38
rs1017887289 Chr.9: 137251685-137251685 on Build GRCh38
rs746830820 Chr.9: 137251773-137251773 on Build GRCh38
rs1031723603 Chr.9: 137251666-137251666 on Build GRCh38
rs774717876 Chr.9: 137251708-137251708 on Build GRCh38
rs1012781310 Chr.9: 137251627-137251627 on Build GRCh38
rs369576826 Chr.9: 137251723-137251724 on Build GRCh38
rs770091146 Chr.9: 137251774-137251774 on Build GRCh38
rs777364709 Chr.9: 137251772-137251772 on Build GRCh38
rs766046335 Chr.9: 137251734-137251734 on Build GRCh38
rs201942432 Chr.9: 137251701-137251701 on Build GRCh38
rs998940944 Chr.9: 137251652-137251652 on Build GRCh38
rs560546642 Chr.9: 137251732-137251732 on Build GRCh38
rs1038290624 Chr.9: 137251585-137251585 on Build GRCh38
rs373392997 Chr.9: 137251759-137251759 on Build GRCh38
rs755727943 Chr.9: 137251723-137251723 on Build GRCh38
rs928664057 Chr.9: 137251576-137251576 on Build GRCh38
rs868403122 Chr.9: 137251577-137251577 on Build GRCh38
rs201778051 Chr.9: 137251777-137251777 on Build GRCh38
rs750792351 Chr.9: 137251722-137251722 on Build GRCh38
rs200313176 Chr.9: 137251767-137251767 on Build GRCh38
rs894365199 Chr.9: 137251601-137251601 on Build GRCh38
rs199944835 Chr.9: 137251768-137251768 on Build GRCh38
rs753541365 Chr.9: 137251744-137251744 on Build GRCh38
rs370217083 Chr.9: 137251697-137251697 on Build GRCh38

View Full Product Details