Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96842159-96842404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD23
Entrez Gene ID
200539
Gene Name
ankyrin repeat domain 23
Gene Aliases
DARP, MARP3
UniGene
Hs.643430
Gene Chromosome Location
Chr.2: 96837914-96844021 on Build GRCh38
Transcripts
NM_144994.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6600474 c.141G>A p.E47E Chr.2: 96842398-96842398 on Build GRCh38
1023999 c.175-1G>A p.? Chr.2: 96842186-96842186 on Build GRCh38
6988466 c.137G>A p.R46Q Chr.2: 96842402-96842402 on Build GRCh38
1024000 c.171_173delGAA p.K58delK Chr.2: 96842366-96842368 on Build GRCh38
4779484 c.150A>G p.K50K Chr.2: 96842389-96842389 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377763483 Chr.2: 96842313-96842313 on Build GRCh38
rs939741820 Chr.2: 96842205-96842205 on Build GRCh38
rs760223637 Chr.2: 96842354-96842354 on Build GRCh38
rs758051429 Chr.2: 96842360-96842363 on Build GRCh38
rs949007391 Chr.2: 96842257-96842257 on Build GRCh38
rs754385371 Chr.2: 96842378-96842380 on Build GRCh38
rs779616716 Chr.2: 96842366-96842368 on Build GRCh38
rs750280463 Chr.2: 96842345-96842346 on Build GRCh38
rs559206940 Chr.2: 96842346-96842346 on Build GRCh38
rs374826545 Chr.2: 96842250-96842250 on Build GRCh38
rs566880489 Chr.2: 96842293-96842293 on Build GRCh38
rs111422274 Chr.2: 96842362-96842362 on Build GRCh38
rs769889678 Chr.2: 96842337-96842337 on Build GRCh38
rs765012867 Chr.2: 96842341-96842346 on Build GRCh38
rs907221387 Chr.2: 96842312-96842312 on Build GRCh38
rs1044626326 Chr.2: 96842300-96842300 on Build GRCh38
rs1038504463 Chr.2: 96842221-96842221 on Build GRCh38
rs34208858 Chr.2: 96842346-96842347 on Build GRCh38
rs766010989 Chr.2: 96842357-96842357 on Build GRCh38
rs993742185 Chr.2: 96842193-96842193 on Build GRCh38
rs745971290 Chr.2: 96842332-96842332 on Build GRCh38
rs781321884 Chr.2: 96842233-96842233 on Build GRCh38
rs575529407 Chr.2: 96842352-96842352 on Build GRCh38
rs143591529 Chr.2: 96842282-96842282 on Build GRCh38
rs1011630452 Chr.2: 96842348-96842348 on Build GRCh38
rs536730749 Chr.2: 96842279-96842279 on Build GRCh38
rs201015005 Chr.2: 96842377-96842377 on Build GRCh38
rs759270586 Chr.2: 96842371-96842371 on Build GRCh38
rs751259312 Chr.2: 96842367-96842377 on Build GRCh38
rs1025686329 Chr.2: 96842264-96842264 on Build GRCh38
rs567802855 Chr.2: 96842274-96842274 on Build GRCh38
rs113375706 Chr.2: 96842364-96842364 on Build GRCh38
rs58147284 Chr.2: 96842345-96842345 on Build GRCh38
rs757773427 Chr.2: 96842213-96842213 on Build GRCh38
rs747487070 Chr.2: 96842209-96842209 on Build GRCh38
rs761794999 Chr.2: 96842351-96842351 on Build GRCh38
rs375900322 Chr.2: 96842344-96842344 on Build GRCh38
rs780197299 Chr.2: 96842341-96842341 on Build GRCh38
rs778300385 Chr.2: 96842207-96842207 on Build GRCh38

View Full Product Details