Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86344498-86344757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIKESHI
Entrez Gene ID
51501
Gene Name
Hikeshi, heat shock protein nuclear import factor
Gene Aliases
C11orf73, HLD13, HSPC138, HSPC179, L7RN6, OPI10
UniGene
Hs.283322
Gene Chromosome Location
Chr.11: 86302211-86345943 on Build GRCh38
Transcripts
NM_001322404.1 NM_001322407.1 NM_001322409.1 NM_016401.3 NR_024597.1 NR_024598.1 NR_136324.1 XM_017017914.1 XM_017017915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
932721 c.518C>T p.P173L Chr.11: 86344700-86344700 on Build GRCh38
279248 c.432G>T p.K144N Chr.11: 86344614-86344614 on Build GRCh38
5839745 c.517C>T p.P173S Chr.11: 86344699-86344699 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs150225885 Chr.11: 86344634-86344634 on Build GRCh38
rs773000365 Chr.11: 86344641-86344641 on Build GRCh38
rs200478904 Chr.11: 86344653-86344653 on Build GRCh38
rs768659728 Chr.11: 86344618-86344618 on Build GRCh38
rs543736607 Chr.11: 86344566-86344566 on Build GRCh38
rs760191235 Chr.11: 86344678-86344678 on Build GRCh38
rs778801770 Chr.11: 86344625-86344625 on Build GRCh38
rs911095529 Chr.11: 86344551-86344551 on Build GRCh38
rs898415189 Chr.11: 86344614-86344614 on Build GRCh38
rs1040377300 Chr.11: 86344587-86344587 on Build GRCh38
rs779946715 Chr.11: 86344579-86344579 on Build GRCh38
rs369926601 Chr.11: 86344574-86344574 on Build GRCh38
rs749414453 Chr.11: 86344602-86344602 on Build GRCh38
rs1132040 Chr.11: 86344722-86344722 on Build GRCh38
rs764170602 Chr.11: 86344714-86344714 on Build GRCh38
rs578131339 Chr.11: 86344701-86344701 on Build GRCh38
rs529918556 Chr.11: 86344565-86344565 on Build GRCh38
rs748141990 Chr.11: 86344630-86344630 on Build GRCh38
rs373031648 Chr.11: 86344696-86344696 on Build GRCh38
rs901839969 Chr.11: 86344598-86344598 on Build GRCh38
rs141742418 Chr.11: 86344716-86344716 on Build GRCh38
rs887955906 Chr.11: 86344690-86344690 on Build GRCh38
rs1003900454 Chr.11: 86344547-86344547 on Build GRCh38
rs759061923 Chr.11: 86344700-86344700 on Build GRCh38
rs985397102 Chr.11: 86344548-86344548 on Build GRCh38
rs775967173 Chr.11: 86344697-86344697 on Build GRCh38
rs368044577 Chr.11: 86344683-86344683 on Build GRCh38
rs764895793 Chr.11: 86344707-86344707 on Build GRCh38

View Full Product Details