Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27055018-27055291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL5
Entrez Gene ID
60509
Gene Name
ATP/GTP binding protein like 5
Gene Aliases
CCP5, RP75
UniGene
Hs.138207
Gene Chromosome Location
Chr.2: 27051423-27070622 on Build GRCh38
Transcripts
NM_001035507.2 NM_021831.5 NR_104246.1 XM_005264477.2 XM_011533011.2 XM_011533012.2 XM_011533013.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7300584 c.883G>A p.D295N Chr.2: 27055228-27055228 on Build GRCh38
4583423 c.842G>A p.R281H Chr.2: 27055187-27055187 on Build GRCh38
6356432 c.853G>A p.V285I Chr.2: 27055198-27055198 on Build GRCh38
2913019 c.841C>T p.R281C Chr.2: 27055186-27055186 on Build GRCh38
4779093 c.814C>A p.P272T Chr.2: 27055159-27055159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766046695 Chr.2: 27055066-27055067 on Build GRCh38
rs780529426 Chr.2: 27055053-27055053 on Build GRCh38
rs772845393 Chr.2: 27055212-27055212 on Build GRCh38
rs373322749 Chr.2: 27055197-27055197 on Build GRCh38
rs750560506 Chr.2: 27055264-27055264 on Build GRCh38
rs768337268 Chr.2: 27055059-27055059 on Build GRCh38
rs759378963 Chr.2: 27055155-27055155 on Build GRCh38
rs766222932 Chr.2: 27055153-27055153 on Build GRCh38
rs879253769 Chr.2: 27055171-27055171 on Build GRCh38
rs757420415 Chr.2: 27055160-27055160 on Build GRCh38
rs17005799 Chr.2: 27055125-27055125 on Build GRCh38
rs759432041 Chr.2: 27055236-27055236 on Build GRCh38
rs774378905 Chr.2: 27055235-27055235 on Build GRCh38
rs747738876 Chr.2: 27055198-27055198 on Build GRCh38
rs141571521 Chr.2: 27055172-27055172 on Build GRCh38
rs769317509 Chr.2: 27055203-27055203 on Build GRCh38
rs764753761 Chr.2: 27055239-27055239 on Build GRCh38
rs763004799 Chr.2: 27055128-27055128 on Build GRCh38
rs762338805 Chr.2: 27055226-27055226 on Build GRCh38
rs778996873 Chr.2: 27055163-27055163 on Build GRCh38
rs557680569 Chr.2: 27055185-27055185 on Build GRCh38
rs867890393 Chr.2: 27055108-27055108 on Build GRCh38
rs759014967 Chr.2: 27055173-27055173 on Build GRCh38
rs150831853 Chr.2: 27055190-27055190 on Build GRCh38
rs948831045 Chr.2: 27055164-27055164 on Build GRCh38
rs370112745 Chr.2: 27055057-27055057 on Build GRCh38
rs776245158 Chr.2: 27055064-27055064 on Build GRCh38
rs764430159 Chr.2: 27055156-27055156 on Build GRCh38
rs772340526 Chr.2: 27055048-27055048 on Build GRCh38
rs746322182 Chr.2: 27055047-27055047 on Build GRCh38
rs762186367 Chr.2: 27055251-27055251 on Build GRCh38
rs780394281 Chr.2: 27055186-27055186 on Build GRCh38
rs537023540 Chr.2: 27055068-27055068 on Build GRCh38
rs375903966 Chr.2: 27055227-27055227 on Build GRCh38
rs879253768 Chr.2: 27055228-27055228 on Build GRCh38
rs202027391 Chr.2: 27055269-27055269 on Build GRCh38
rs754070994 Chr.2: 27055157-27055157 on Build GRCh38
rs887824792 Chr.2: 27055252-27055252 on Build GRCh38
rs990455103 Chr.2: 27055049-27055049 on Build GRCh38
rs373166235 Chr.2: 27055271-27055271 on Build GRCh38
rs754128307 Chr.2: 27055241-27055241 on Build GRCh38
rs369068296 Chr.2: 27055253-27055253 on Build GRCh38
rs868031509 Chr.2: 27055189-27055189 on Build GRCh38
rs769255452 Chr.2: 27055088-27055088 on Build GRCh38

View Full Product Details