Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45183275-45183548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADARB1
Entrez Gene ID
104
Gene Name
adenosine deaminase, RNA specific B1
Gene Aliases
ADAR2, DRABA2, DRADA2, RED1
UniGene
Hs.474018
Gene Chromosome Location
Chr.21: 45074578-45226563 on Build GRCh38
Transcripts
NM_001112.3 NM_001160230.1 NM_015833.3 NM_015834.3 NR_027672.1 NR_027673.1 NR_027674.1 NR_073200.1 XM_006723954.2 XM_011529425.2 XM_011529432.1 XM_017028242.1 XM_017028243.1 XM_017028244.1 XM_017028245.1 XM_017028246.1 XM_017028247.1 XM_017028248.1 XM_017028249.1 XM_017028250.1 XM_017028251.1 XM_017028252.1 XM_017028253.1 XM_017028254.1 XM_017028255.1 XM_017028256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1414477 c.1385C>T p.P462L Chr.21: 45183499-45183499 on Build GRCh38
6304416 c.1292G>A p.R431Q Chr.21: 45183406-45183406 on Build GRCh38
1682047 c.1309A>G p.K437E Chr.21: 45183423-45183423 on Build GRCh38
4973213 c.1286C>T p.S429L Chr.21: 45183400-45183400 on Build GRCh38
6594682 c.1333T>C p.Y445H Chr.21: 45183447-45183447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138735662 Chr.21: 45183471-45183471 on Build GRCh38
rs945417875 Chr.21: 45183410-45183410 on Build GRCh38
rs760565860 Chr.21: 45183393-45183393 on Build GRCh38
rs542356039 Chr.21: 45183520-45183520 on Build GRCh38
rs142574659 Chr.21: 45183398-45183398 on Build GRCh38
rs770828533 Chr.21: 45183334-45183334 on Build GRCh38
rs774090998 Chr.21: 45183341-45183341 on Build GRCh38
rs367739860 Chr.21: 45183318-45183318 on Build GRCh38
rs759967310 Chr.21: 45183326-45183326 on Build GRCh38
rs776820750 Chr.21: 45183323-45183323 on Build GRCh38
rs749940638 Chr.21: 45183387-45183387 on Build GRCh38
rs201189520 Chr.21: 45183522-45183522 on Build GRCh38
rs536652455 Chr.21: 45183355-45183355 on Build GRCh38
rs775659792 Chr.21: 45183372-45183372 on Build GRCh38
rs150188818 Chr.21: 45183419-45183419 on Build GRCh38
rs753019271 Chr.21: 45183467-45183467 on Build GRCh38
rs954570151 Chr.21: 45183380-45183380 on Build GRCh38
rs267606164 Chr.21: 45183503-45183503 on Build GRCh38
rs758704261 Chr.21: 45183501-45183501 on Build GRCh38
rs779000758 Chr.21: 45183458-45183458 on Build GRCh38
rs754844391 Chr.21: 45183408-45183408 on Build GRCh38
rs139637757 Chr.21: 45183386-45183386 on Build GRCh38
rs1030194940 Chr.21: 45183347-45183347 on Build GRCh38
rs753556817 Chr.21: 45183406-45183406 on Build GRCh38
rs747112715 Chr.21: 45183519-45183519 on Build GRCh38
rs372108954 Chr.21: 45183514-45183514 on Build GRCh38

View Full Product Details