Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58351318-58351591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A1BG
Entrez Gene ID
1
Gene Name
alpha-1-B glycoprotein
Gene Aliases
A1B, ABG, GAB, HYST2477
UniGene
Hs.529161
Gene Chromosome Location
Chr.19: 58346806-58353499 on Build GRCh38
Transcripts
NM_130786.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5959904 c.711T>G p.S237R Chr.19: 58351590-58351590 on Build GRCh38
4557179 c.717G>A p.V239V Chr.19: 58351584-58351584 on Build GRCh38
4597446 c.812T>G p.L271R Chr.19: 58351489-58351489 on Build GRCh38
2974308 c.770C>A p.T257N Chr.19: 58351531-58351531 on Build GRCh38
6589080 c.863delA p.N288fs*12 Chr.19: 58351438-58351438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145717206 Chr.19: 58351509-58351509 on Build GRCh38
rs930140223 Chr.19: 58351559-58351559 on Build GRCh38
rs1050564984 Chr.19: 58351482-58351482 on Build GRCh38
rs143384169 Chr.19: 58351424-58351424 on Build GRCh38
rs777113231 Chr.19: 58351525-58351525 on Build GRCh38
rs759201192 Chr.19: 58351541-58351541 on Build GRCh38
rs775736468 Chr.19: 58351551-58351551 on Build GRCh38
rs746361920 Chr.19: 58351527-58351527 on Build GRCh38
rs376793710 Chr.19: 58351361-58351361 on Build GRCh38
rs200002683 Chr.19: 58351565-58351565 on Build GRCh38
rs746494365 Chr.19: 58351515-58351515 on Build GRCh38
rs780117454 Chr.19: 58351495-58351495 on Build GRCh38
rs761363571 Chr.19: 58351407-58351407 on Build GRCh38
rs149231769 Chr.19: 58351417-58351417 on Build GRCh38
rs778614779 Chr.19: 58351457-58351457 on Build GRCh38
rs770480372 Chr.19: 58351520-58351520 on Build GRCh38
rs368461257 Chr.19: 58351494-58351494 on Build GRCh38
rs776096132 Chr.19: 58351539-58351539 on Build GRCh38
rs773913887 Chr.19: 58351400-58351400 on Build GRCh38
rs200592383 Chr.19: 58351469-58351469 on Build GRCh38
rs375770905 Chr.19: 58351468-58351468 on Build GRCh38
rs371831204 Chr.19: 58351498-58351498 on Build GRCh38
rs768385007 Chr.19: 58351394-58351394 on Build GRCh38
rs747779138 Chr.19: 58351503-58351503 on Build GRCh38
rs771996787 Chr.19: 58351341-58351342 on Build GRCh38
rs777166096 Chr.19: 58351471-58351471 on Build GRCh38
rs771544362 Chr.19: 58351467-58351467 on Build GRCh38
rs747822956 Chr.19: 58351462-58351462 on Build GRCh38
rs371502566 Chr.19: 58351358-58351358 on Build GRCh38
rs771541338 Chr.19: 58351410-58351410 on Build GRCh38
rs777715904 Chr.19: 58351430-58351430 on Build GRCh38
rs1057280764 Chr.19: 58351341-58351341 on Build GRCh38
rs145532647 Chr.19: 58351571-58351571 on Build GRCh38
rs773932487 Chr.19: 58351360-58351360 on Build GRCh38
rs771461441 Chr.19: 58351470-58351470 on Build GRCh38
rs778812426 Chr.19: 58351375-58351375 on Build GRCh38
rs80353491 Chr.19: 58351570-58351570 on Build GRCh38
rs1047140142 Chr.19: 58351374-58351374 on Build GRCh38
rs770134003 Chr.19: 58351537-58351537 on Build GRCh38
rs749058804 Chr.19: 58351442-58351442 on Build GRCh38
rs756458167 Chr.19: 58351367-58351367 on Build GRCh38
rs755207686 Chr.19: 58351371-58351371 on Build GRCh38
rs765582780 Chr.19: 58351474-58351474 on Build GRCh38
rs773705977 Chr.19: 58351411-58351411 on Build GRCh38
rs753884034 Chr.19: 58351497-58351497 on Build GRCh38
rs781171705 Chr.19: 58351383-58351383 on Build GRCh38
rs758495858 Chr.19: 58351428-58351428 on Build GRCh38
rs780004935 Chr.19: 58351387-58351387 on Build GRCh38
rs527508602 Chr.19: 58351508-58351508 on Build GRCh38
rs750325654 Chr.19: 58351431-58351431 on Build GRCh38
rs372443839 Chr.19: 58351368-58351368 on Build GRCh38
rs147717500 Chr.19: 58351413-58351413 on Build GRCh38
rs542119157 Chr.19: 58351414-58351414 on Build GRCh38
rs192588134 Chr.19: 58351379-58351379 on Build GRCh38
rs765941252 Chr.19: 58351499-58351499 on Build GRCh38
rs775274688 Chr.19: 58351349-58351349 on Build GRCh38
rs773194551 Chr.19: 58351345-58351345 on Build GRCh38
rs545512616 Chr.19: 58351363-58351363 on Build GRCh38
rs889573031 Chr.19: 58351489-58351489 on Build GRCh38
rs763192368 Chr.19: 58351553-58351553 on Build GRCh38
rs756140080 Chr.19: 58351436-58351436 on Build GRCh38
rs759926508 Chr.19: 58351472-58351472 on Build GRCh38
rs760124077 Chr.19: 58351420-58351421 on Build GRCh38
rs968803656 Chr.19: 58351437-58351437 on Build GRCh38
rs778577569 Chr.19: 58351500-58351500 on Build GRCh38
rs1010426141 Chr.19: 58351418-58351418 on Build GRCh38
rs576303734 Chr.19: 58351384-58351384 on Build GRCh38
rs763094323 Chr.19: 58351477-58351477 on Build GRCh38
rs151257234 Chr.19: 58351456-58351456 on Build GRCh38
rs746222025 Chr.19: 58351339-58351339 on Build GRCh38
rs768035808 Chr.19: 58351362-58351362 on Build GRCh38
rs377700571 Chr.19: 58351385-58351385 on Build GRCh38
rs764378776 Chr.19: 58351564-58351564 on Build GRCh38
rs868157705 Chr.19: 58351463-58351463 on Build GRCh38
rs184830744 Chr.19: 58351568-58351568 on Build GRCh38
rs140428520 Chr.19: 58351491-58351491 on Build GRCh38
rs371215771 Chr.19: 58351392-58351392 on Build GRCh38
rs754798923 Chr.19: 58351499-58351499 on Build GRCh38
rs769765377 Chr.19: 58351348-58351348 on Build GRCh38
rs753106272 Chr.19: 58351562-58351563 on Build GRCh38
rs973669068 Chr.19: 58351382-58351382 on Build GRCh38
rs765469947 Chr.19: 58351419-58351419 on Build GRCh38
rs779953572 Chr.19: 58351440-58351440 on Build GRCh38
rs754226646 Chr.19: 58351416-58351416 on Build GRCh38
rs758452143 Chr.19: 58351381-58351381 on Build GRCh38
rs374586221 Chr.19: 58351566-58351566 on Build GRCh38
rs753993883 Chr.19: 58351369-58351369 on Build GRCh38
rs764833418 Chr.19: 58351550-58351550 on Build GRCh38

View Full Product Details