Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10430997-10431252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7IP2
Entrez Gene ID
80063
Gene Name
activating transcription factor 7 interacting protein 2
Gene Aliases
MCAF2
UniGene
Hs.513343
Gene Chromosome Location
Chr.16: 10386055-10483640 on Build GRCh38
Transcripts
NM_001256160.1 NM_024997.3 NR_045815.1 NR_045816.1 XM_006720953.3 XM_006720954.3 XM_017023705.1 XM_017023706.1 XM_017023707.1 XM_017023710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4663991 c.504C>T p.C168C Chr.16: 10431124-10431124 on Build GRCh38
4986749 c.446C>T p.T149I Chr.16: 10431066-10431066 on Build GRCh38
3887858 c.576G>A p.K192K Chr.16: 10431196-10431196 on Build GRCh38
3887857 c.442C>T p.Q148* Chr.16: 10431062-10431062 on Build GRCh38
3505696 c.461C>T p.S154F Chr.16: 10431081-10431081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202209292 Chr.16: 10431184-10431184 on Build GRCh38
rs775862931 Chr.16: 10431219-10431219 on Build GRCh38
rs766329838 Chr.16: 10431163-10431163 on Build GRCh38
rs542238830 Chr.16: 10431183-10431183 on Build GRCh38
rs755012666 Chr.16: 10431177-10431177 on Build GRCh38
rs76697507 Chr.16: 10431097-10431097 on Build GRCh38
rs746426429 Chr.16: 10431119-10431119 on Build GRCh38
rs752195575 Chr.16: 10431179-10431179 on Build GRCh38
rs758269231 Chr.16: 10431068-10431068 on Build GRCh38
rs145957232 Chr.16: 10431101-10431101 on Build GRCh38
rs760681694 Chr.16: 10431132-10431132 on Build GRCh38
rs767685356 Chr.16: 10431038-10431038 on Build GRCh38
rs563342232 Chr.16: 10431148-10431150 on Build GRCh38
rs774843896 Chr.16: 10431020-10431020 on Build GRCh38
rs764925383 Chr.16: 10431056-10431056 on Build GRCh38
rs771575139 Chr.16: 10431050-10431050 on Build GRCh38
rs369166200 Chr.16: 10431185-10431185 on Build GRCh38
rs760006935 Chr.16: 10431147-10431147 on Build GRCh38
rs751515922 Chr.16: 10431171-10431171 on Build GRCh38
rs751586601 Chr.16: 10431187-10431187 on Build GRCh38
rs766374861 Chr.16: 10431071-10431071 on Build GRCh38
rs1031043183 Chr.16: 10431110-10431110 on Build GRCh38
rs759494436 Chr.16: 10431031-10431031 on Build GRCh38
rs534075576 Chr.16: 10431063-10431063 on Build GRCh38
rs74322853 Chr.16: 10431150-10431150 on Build GRCh38
rs941001889 Chr.16: 10431210-10431210 on Build GRCh38
rs34430103 Chr.16: 10431100-10431100 on Build GRCh38
rs775695051 Chr.16: 10431129-10431129 on Build GRCh38
rs757586127 Chr.16: 10431199-10431199 on Build GRCh38
rs774817108 Chr.16: 10431117-10431117 on Build GRCh38
rs114568129 Chr.16: 10431049-10431049 on Build GRCh38
rs377208400 Chr.16: 10431087-10431087 on Build GRCh38
rs777021803 Chr.16: 10431142-10431142 on Build GRCh38
rs966561237 Chr.16: 10431121-10431121 on Build GRCh38
rs150784930 Chr.16: 10431092-10431092 on Build GRCh38
rs771316238 Chr.16: 10431102-10431102 on Build GRCh38
rs980170212 Chr.16: 10431022-10431022 on Build GRCh38
rs118085445 Chr.16: 10431205-10431205 on Build GRCh38
rs541287771 Chr.16: 10431217-10431217 on Build GRCh38
rs772038491 Chr.16: 10431120-10431120 on Build GRCh38
rs999506844 Chr.16: 10431107-10431107 on Build GRCh38
rs371774821 Chr.16: 10431178-10431178 on Build GRCh38
rs779317235 Chr.16: 10431203-10431203 on Build GRCh38
rs764342084 Chr.16: 10431135-10431135 on Build GRCh38
rs1029873196 Chr.16: 10431151-10431151 on Build GRCh38

View Full Product Details