Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 25468860-25469117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8A2
Entrez Gene ID
51761
Gene Name
ATPase phospholipid transporting 8A2
Gene Aliases
ATP, ATPIB, CAMRQ4, IB, ML-1
UniGene
Hs.444957
Gene Chromosome Location
Chr.13: 25372011-26025851 on Build GRCh38
Transcripts
NM_001313741.1 NM_016529.5 XM_005266419.1 XM_011535103.1 XM_011535104.2 XM_011535106.1 XM_011535107.2 XM_011535113.2 XM_017020625.1 XM_017020626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6608277 c.22G>T p.G8* Chr.13: 25469042-25469042 on Build GRCh38
4527364 c.25G>A p.D9N Chr.13: 25469045-25469045 on Build GRCh38
4046757 c.158C>T p.A53V Chr.13: 25469058-25469058 on Build GRCh38
1366101 c.144A>C p.G48G Chr.13: 25469044-25469044 on Build GRCh38
5639442 c.165C>T p.A55A Chr.13: 25469065-25469065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992751398 Chr.13: 25469029-25469029 on Build GRCh38
rs7317185 Chr.13: 25469044-25469044 on Build GRCh38
rs759505168 Chr.13: 25468939-25468939 on Build GRCh38
rs928463041 Chr.13: 25468880-25468880 on Build GRCh38
rs373909734 Chr.13: 25468975-25468975 on Build GRCh38
rs775292632 Chr.13: 25469069-25469069 on Build GRCh38
rs12872195 Chr.13: 25469096-25469096 on Build GRCh38
rs376409652 Chr.13: 25468949-25468949 on Build GRCh38
rs1056157223 Chr.13: 25468892-25468892 on Build GRCh38
rs749986570 Chr.13: 25468973-25468973 on Build GRCh38
rs864309608 Chr.13: 25469045-25469045 on Build GRCh38
rs982197445 Chr.13: 25469089-25469089 on Build GRCh38
rs774959381 Chr.13: 25469030-25469031 on Build GRCh38
rs1025087733 Chr.13: 25469035-25469035 on Build GRCh38
rs761517856 Chr.13: 25469084-25469084 on Build GRCh38
rs756806400 Chr.13: 25468971-25468971 on Build GRCh38
rs775304354 Chr.13: 25468947-25468947 on Build GRCh38
rs769713590 Chr.13: 25469064-25469064 on Build GRCh38
rs1018303994 Chr.13: 25469025-25469025 on Build GRCh38
rs202073376 Chr.13: 25469058-25469058 on Build GRCh38
rs377512165 Chr.13: 25469063-25469063 on Build GRCh38
rs932480898 Chr.13: 25468884-25468884 on Build GRCh38
rs1009468564 Chr.13: 25468916-25468916 on Build GRCh38
rs12875883 Chr.13: 25469092-25469092 on Build GRCh38
rs938492112 Chr.13: 25468890-25468890 on Build GRCh38
rs199807614 Chr.13: 25468959-25468959 on Build GRCh38
rs200308244 Chr.13: 25468929-25468929 on Build GRCh38
rs545159626 Chr.13: 25469076-25469076 on Build GRCh38
rs373863313 Chr.13: 25468896-25468896 on Build GRCh38
rs12875744 Chr.13: 25469040-25469040 on Build GRCh38
rs767176234 Chr.13: 25468972-25468972 on Build GRCh38
rs755369347 Chr.13: 25469026-25469026 on Build GRCh38
rs765115743 Chr.13: 25468945-25468945 on Build GRCh38
rs762803460 Chr.13: 25469082-25469082 on Build GRCh38
rs748413967 Chr.13: 25469047-25469047 on Build GRCh38
rs959460010 Chr.13: 25469003-25469003 on Build GRCh38
rs767302786 Chr.13: 25469086-25469086 on Build GRCh38
rs201409188 Chr.13: 25468977-25468977 on Build GRCh38
rs539394433 Chr.13: 25468898-25468898 on Build GRCh38
rs573165206 Chr.13: 25468956-25468956 on Build GRCh38
rs1037388289 Chr.13: 25468899-25468899 on Build GRCh38
rs749896642 Chr.13: 25469090-25469090 on Build GRCh38
rs370672539 Chr.13: 25469083-25469083 on Build GRCh38

View Full Product Details