Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 10700128-10700369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf74
Entrez Gene ID
203076
Gene Name
chromosome 8 open reading frame 74
Gene Aliases
-
UniGene
Hs.371776
Gene Chromosome Location
Chr.8: 10672557-10700616 on Build GRCh38
Transcripts
NM_001040032.1 XM_011543816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6258074 c.729C>A p.N243K Chr.8: 10700315-10700315 on Build GRCh38
6258073 c.687G>T p.Q229H Chr.8: 10700273-10700273 on Build GRCh38
1094666 c.759G>T p.Q253H Chr.8: 10700345-10700345 on Build GRCh38
1283851 c.699G>T p.L233L Chr.8: 10700285-10700285 on Build GRCh38
4508420 c.775C>T p.L259F Chr.8: 10700361-10700361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759700922 Chr.8: 10700297-10700297 on Build GRCh38
rs371469734 Chr.8: 10700321-10700321 on Build GRCh38
rs374916501 Chr.8: 10700322-10700322 on Build GRCh38
rs769860984 Chr.8: 10700216-10700216 on Build GRCh38
rs761170615 Chr.8: 10700284-10700284 on Build GRCh38
rs907583003 Chr.8: 10700254-10700254 on Build GRCh38
rs765461161 Chr.8: 10700301-10700301 on Build GRCh38
rs764264532 Chr.8: 10700315-10700315 on Build GRCh38
rs752775876 Chr.8: 10700233-10700233 on Build GRCh38
rs961924312 Chr.8: 10700151-10700151 on Build GRCh38
rs763068879 Chr.8: 10700304-10700304 on Build GRCh38
rs565388774 Chr.8: 10700318-10700318 on Build GRCh38
rs751194554 Chr.8: 10700175-10700176 on Build GRCh38
rs1054421610 Chr.8: 10700220-10700220 on Build GRCh38
rs535256632 Chr.8: 10700282-10700282 on Build GRCh38
rs778366643 Chr.8: 10700333-10700333 on Build GRCh38
rs117295315 Chr.8: 10700207-10700207 on Build GRCh38
rs563141070 Chr.8: 10700187-10700187 on Build GRCh38
rs375171655 Chr.8: 10700237-10700237 on Build GRCh38
rs747567366 Chr.8: 10700334-10700334 on Build GRCh38
rs769437570 Chr.8: 10700270-10700270 on Build GRCh38
rs1021511174 Chr.8: 10700219-10700219 on Build GRCh38
rs200464332 Chr.8: 10700245-10700245 on Build GRCh38
rs892473238 Chr.8: 10700330-10700330 on Build GRCh38
rs1052517891 Chr.8: 10700324-10700324 on Build GRCh38
rs201803874 Chr.8: 10700213-10700213 on Build GRCh38
rs374602916 Chr.8: 10700253-10700253 on Build GRCh38
rs745555031 Chr.8: 10700269-10700269 on Build GRCh38
rs57041981 Chr.8: 10700250-10700250 on Build GRCh38
rs374346316 Chr.8: 10700266-10700266 on Build GRCh38
rs980311476 Chr.8: 10700232-10700232 on Build GRCh38
rs751586132 Chr.8: 10700316-10700316 on Build GRCh38
rs780870166 Chr.8: 10700320-10700320 on Build GRCh38
rs771243175 Chr.8: 10700287-10700287 on Build GRCh38
rs781008020 Chr.8: 10700267-10700267 on Build GRCh38
rs186456612 Chr.8: 10700152-10700152 on Build GRCh38
rs755194433 Chr.8: 10700221-10700221 on Build GRCh38
rs765279424 Chr.8: 10700222-10700222 on Build GRCh38
rs148093950 Chr.8: 10700190-10700190 on Build GRCh38
rs1014579384 Chr.8: 10700149-10700149 on Build GRCh38
rs140949609 Chr.8: 10700194-10700194 on Build GRCh38
rs953305170 Chr.8: 10700167-10700167 on Build GRCh38
rs879194116 Chr.8: 10700293-10700293 on Build GRCh38
rs768226569 Chr.8: 10700331-10700331 on Build GRCh38
rs760976050 Chr.8: 10700214-10700214 on Build GRCh38
rs781272422 Chr.8: 10700164-10700164 on Build GRCh38
rs569861669 Chr.8: 10700260-10700260 on Build GRCh38
rs370382914 Chr.8: 10700217-10700217 on Build GRCh38
rs748726419 Chr.8: 10700277-10700277 on Build GRCh38
rs116653093 Chr.8: 10700211-10700211 on Build GRCh38
rs191237807 Chr.8: 10700191-10700191 on Build GRCh38
rs965492677 Chr.8: 10700202-10700202 on Build GRCh38
rs746759282 Chr.8: 10700188-10700188 on Build GRCh38
rs774891538 Chr.8: 10700274-10700274 on Build GRCh38
rs777034726 Chr.8: 10700294-10700294 on Build GRCh38
rs368576265 Chr.8: 10700302-10700302 on Build GRCh38
rs932183433 Chr.8: 10700288-10700288 on Build GRCh38
rs897474308 Chr.8: 10700283-10700283 on Build GRCh38

View Full Product Details