Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150330683-150330946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC61
Entrez Gene ID
65999
Gene Name
leucine rich repeat containing 61
Gene Aliases
HSPC295
UniGene
Hs.647119
Gene Chromosome Location
Chr.7: 150322639-150338156 on Build GRCh38
Transcripts
NM_001142928.1 NM_023942.2 XM_006716095.2 NM_138434.2

Gene Symbol
ZBED6CL
Entrez Gene ID
113763
Gene Name
ZBED6 C-terminal like
Gene Aliases
C7orf29
UniGene
Hs.655915
Gene Chromosome Location
Chr.7: 150329849-150332721 on Build GRCh38
Transcripts
NM_001142928.1 NM_023942.2 XM_006716095.2 NM_138434.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
73964 c.310G>C p.V104L Chr.7: 150330714-150330714 on Build GRCh38
3923067 c.427G>A p.E143K Chr.7: 150330831-150330831 on Build GRCh38
5552855 c.446A>G p.E149G Chr.7: 150330850-150330850 on Build GRCh38
5558734 c.510G>A p.L170L Chr.7: 150330914-150330914 on Build GRCh38
5163105 c.428A>C p.E143A Chr.7: 150330832-150330832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898255761 Chr.7: 150330880-150330880 on Build GRCh38
rs201290942 Chr.7: 150330740-150330740 on Build GRCh38
rs780657679 Chr.7: 150330784-150330784 on Build GRCh38
rs769154561 Chr.7: 150330842-150330842 on Build GRCh38
rs767074213 Chr.7: 150330771-150330771 on Build GRCh38
rs781085130 Chr.7: 150330719-150330719 on Build GRCh38
rs754775165 Chr.7: 150330794-150330794 on Build GRCh38
rs1029094149 Chr.7: 150330852-150330852 on Build GRCh38
rs749453935 Chr.7: 150330841-150330841 on Build GRCh38
rs769352786 Chr.7: 150330755-150330755 on Build GRCh38
rs781373331 Chr.7: 150330822-150330822 on Build GRCh38
rs752405524 Chr.7: 150330887-150330887 on Build GRCh38
rs556643007 Chr.7: 150330795-150330795 on Build GRCh38
rs151192485 Chr.7: 150330897-150330897 on Build GRCh38
rs753595180 Chr.7: 150330710-150330710 on Build GRCh38
rs374205555 Chr.7: 150330714-150330714 on Build GRCh38
rs772424036 Chr.7: 150330853-150330853 on Build GRCh38
rs889924476 Chr.7: 150330891-150330891 on Build GRCh38
rs142223774 Chr.7: 150330867-150330867 on Build GRCh38
rs754132303 Chr.7: 150330920-150330920 on Build GRCh38
rs143629316 Chr.7: 150330753-150330753 on Build GRCh38
rs759143074 Chr.7: 150330873-150330873 on Build GRCh38
rs143410668 Chr.7: 150330757-150330757 on Build GRCh38
rs749956364 Chr.7: 150330804-150330804 on Build GRCh38
rs779371947 Chr.7: 150330926-150330926 on Build GRCh38
rs770365616 Chr.7: 150330865-150330865 on Build GRCh38
rs759681404 Chr.7: 150330824-150330824 on Build GRCh38
rs760177664 Chr.7: 150330863-150330863 on Build GRCh38
rs996652471 Chr.7: 150330844-150330844 on Build GRCh38
rs553249346 Chr.7: 150330829-150330829 on Build GRCh38
rs116457847 Chr.7: 150330809-150330809 on Build GRCh38
rs573584543 Chr.7: 150330808-150330808 on Build GRCh38
rs1036773432 Chr.7: 150330875-150330875 on Build GRCh38
rs766007281 Chr.7: 150330787-150330787 on Build GRCh38
rs536790669 Chr.7: 150330781-150330781 on Build GRCh38
rs957225603 Chr.7: 150330784-150330784 on Build GRCh38
rs766189677 Chr.7: 150330703-150330703 on Build GRCh38
rs780418320 Chr.7: 150330838-150330838 on Build GRCh38
rs780223349 Chr.7: 150330911-150330911 on Build GRCh38
rs147992251 Chr.7: 150330844-150330845 on Build GRCh38
rs756264942 Chr.7: 150330898-150330898 on Build GRCh38
rs746134193 Chr.7: 150330733-150330733 on Build GRCh38
rs139910953 Chr.7: 150330749-150330749 on Build GRCh38
rs772554174 Chr.7: 150330773-150330773 on Build GRCh38
rs748717770 Chr.7: 150330845-150330845 on Build GRCh38
rs756462889 Chr.7: 150330836-150330836 on Build GRCh38
rs111533398 Chr.7: 150330716-150330716 on Build GRCh38
rs948677470 Chr.7: 150330823-150330823 on Build GRCh38
rs779096509 Chr.7: 150330844-150330844 on Build GRCh38
rs75555338 Chr.7: 150330713-150330713 on Build GRCh38
rs762998466 Chr.7: 150330889-150330889 on Build GRCh38
rs762167091 Chr.7: 150330763-150330763 on Build GRCh38
rs371370835 Chr.7: 150330921-150330921 on Build GRCh38
rs763925605 Chr.7: 150330890-150330890 on Build GRCh38
rs201560617 Chr.7: 150330857-150330857 on Build GRCh38
rs1016049931 Chr.7: 150330808-150330808 on Build GRCh38

View Full Product Details