Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 4846917-4847190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1E2
Entrez Gene ID
83592
Gene Name
aldo-keto reductase family 1 member E2
Gene Aliases
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR
UniGene
Hs.657944
Gene Chromosome Location
Chr.10: 4824554-4908421 on Build GRCh38
Transcripts
NM_001040177.2 NM_001271021.1 NM_001271025.1 NR_073125.1 NR_073126.1 NR_073127.1 XM_011519715.2 XM_011519718.2 XM_011519720.2 XM_011519722.2 XM_011519724.2 XM_017016744.1 XM_017016745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182436 c.838-1G>T p.? Chr.10: 4847147-4847147 on Build GRCh38
5370679 c.852A>T p.E284D Chr.10: 4847162-4847162 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1052693653 Chr.10: 4847035-4847035 on Build GRCh38
rs548727572 Chr.10: 4847160-4847160 on Build GRCh38
rs532876348 Chr.10: 4847006-4847006 on Build GRCh38
rs112339400 Chr.10: 4847108-4847108 on Build GRCh38
rs758880500 Chr.10: 4847128-4847128 on Build GRCh38
rs909540739 Chr.10: 4846979-4846979 on Build GRCh38
rs753023023 Chr.10: 4847125-4847125 on Build GRCh38
rs12571543 Chr.10: 4847073-4847073 on Build GRCh38
rs35821773 Chr.10: 4847043-4847044 on Build GRCh38
rs904626072 Chr.10: 4847080-4847080 on Build GRCh38
rs1003493897 Chr.10: 4847013-4847013 on Build GRCh38
rs892327930 Chr.10: 4847066-4847066 on Build GRCh38
rs1037232820 Chr.10: 4847092-4847092 on Build GRCh38
rs765594376 Chr.10: 4847121-4847121 on Build GRCh38
rs17133762 Chr.10: 4846947-4846947 on Build GRCh38
rs567336778 Chr.10: 4847140-4847140 on Build GRCh38
rs1027449724 Chr.10: 4847144-4847144 on Build GRCh38
rs1003111392 Chr.10: 4847085-4847085 on Build GRCh38
rs762945353 Chr.10: 4847008-4847008 on Build GRCh38
rs923759541 Chr.10: 4846953-4846953 on Build GRCh38
rs906406689 Chr.10: 4847009-4847009 on Build GRCh38
rs549728803 Chr.10: 4846946-4846946 on Build GRCh38
rs200567525 Chr.10: 4847115-4847115 on Build GRCh38
rs947697564 Chr.10: 4846996-4846996 on Build GRCh38
rs59470948 Chr.10: 4847069-4847069 on Build GRCh38
rs936468146 Chr.10: 4846976-4846976 on Build GRCh38
rs897312507 Chr.10: 4847096-4847096 on Build GRCh38
rs747020328 Chr.10: 4847126-4847126 on Build GRCh38
rs760281299 Chr.10: 4847116-4847116 on Build GRCh38
rs866613502 Chr.10: 4846974-4846974 on Build GRCh38
rs995607005 Chr.10: 4847141-4847141 on Build GRCh38
rs370909662 Chr.10: 4847124-4847124 on Build GRCh38
rs1053944361 Chr.10: 4846978-4846978 on Build GRCh38
rs778256002 Chr.10: 4847130-4847130 on Build GRCh38
rs1048157757 Chr.10: 4846950-4846950 on Build GRCh38
rs947669367 Chr.10: 4847020-4847020 on Build GRCh38
rs752126529 Chr.10: 4847137-4847137 on Build GRCh38
rs536798668 Chr.10: 4847089-4847089 on Build GRCh38
rs781298195 Chr.10: 4847164-4847164 on Build GRCh38

View Full Product Details