Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 45270401-45270628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO6
Entrez Gene ID
196527
Gene Name
anoctamin 6
Gene Aliases
BDPLT7, SCTS, TMEM16F
UniGene
Hs.505339
Gene Chromosome Location
Chr.12: 45215987-45440404 on Build GRCh38
Transcripts
NM_001025356.2 NM_001142679.1 NM_001204803.1 XM_005268707.3

SNP Detail

SNP ID Chromosome Location
rs778562075 Chr.12: 45270590-45270590 on Build GRCh38
rs535887014 Chr.12: 45270497-45270497 on Build GRCh38
rs1027244314 Chr.12: 45270491-45270491 on Build GRCh38
rs746336726 Chr.12: 45270450-45270450 on Build GRCh38
rs1016752613 Chr.12: 45270456-45270456 on Build GRCh38
rs1035375200 Chr.12: 45270561-45270561 on Build GRCh38
rs959769865 Chr.12: 45270591-45270591 on Build GRCh38
rs768914071 Chr.12: 45270539-45270539 on Build GRCh38
rs547786539 Chr.12: 45270467-45270467 on Build GRCh38
rs749638435 Chr.12: 45270512-45270512 on Build GRCh38
rs1031675906 Chr.12: 45270592-45270592 on Build GRCh38
rs899646130 Chr.12: 45270462-45270462 on Build GRCh38
rs780533610 Chr.12: 45270452-45270452 on Build GRCh38
rs977423991 Chr.12: 45270583-45270583 on Build GRCh38
rs780015328 Chr.12: 45270554-45270554 on Build GRCh38
rs982515560 Chr.12: 45270543-45270543 on Build GRCh38
rs772504945 Chr.12: 45270535-45270536 on Build GRCh38
rs995430120 Chr.12: 45270469-45270469 on Build GRCh38
rs10748415 Chr.12: 45270430-45270430 on Build GRCh38
rs1030007484 Chr.12: 45270538-45270538 on Build GRCh38
rs955285642 Chr.12: 45270579-45270579 on Build GRCh38

View Full Product Details