Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36540094-36540361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOAH
Entrez Gene ID
313
Gene Name
acyloxyacyl hydrolase
Gene Aliases
-
UniGene
Hs.488007
Gene Chromosome Location
Chr.7: 36512042-36724549 on Build GRCh38
Transcripts
NM_001177506.1 NM_001177507.1 NM_001637.3 XM_011515333.2 XM_011515334.2 XM_011515335.2 XM_011515336.2 XM_011515338.2 XM_011515339.2 XM_011515340.2 XM_011515341.2 XM_011515342.2 XM_017012102.1 XM_017012103.1 XM_017012104.1 XM_017012105.1 XM_017012106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1089395 c.1300C>T p.P434S Chr.7: 36540325-36540325 on Build GRCh38
3228921 c.1305C>T p.L435L Chr.7: 36540320-36540320 on Build GRCh38
601191 c.1306+1G>A p.? Chr.7: 36540318-36540318 on Build GRCh38
6863020 c.1304T>A p.L435H Chr.7: 36540321-36540321 on Build GRCh38
5718636 c.1292G>A p.R431K Chr.7: 36540333-36540333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776493271 Chr.7: 36540172-36540172 on Build GRCh38
rs772029792 Chr.7: 36540332-36540332 on Build GRCh38
rs979362222 Chr.7: 36540289-36540289 on Build GRCh38
rs142768342 Chr.7: 36540203-36540203 on Build GRCh38
rs965785729 Chr.7: 36540267-36540267 on Build GRCh38
rs774251193 Chr.7: 36540319-36540319 on Build GRCh38
rs566111305 Chr.7: 36540257-36540257 on Build GRCh38
rs772881880 Chr.7: 36540268-36540268 on Build GRCh38
rs533798785 Chr.7: 36540272-36540272 on Build GRCh38
rs1029681942 Chr.7: 36540320-36540320 on Build GRCh38
rs1027847529 Chr.7: 36540180-36540180 on Build GRCh38
rs146048519 Chr.7: 36540312-36540312 on Build GRCh38
rs991906069 Chr.7: 36540225-36540225 on Build GRCh38
rs1024514063 Chr.7: 36540130-36540130 on Build GRCh38
rs769990632 Chr.7: 36540282-36540282 on Build GRCh38
rs967969061 Chr.7: 36540139-36540139 on Build GRCh38
rs576928103 Chr.7: 36540303-36540303 on Build GRCh38
rs374743800 Chr.7: 36540134-36540144 on Build GRCh38
rs140099559 Chr.7: 36540333-36540333 on Build GRCh38
rs1053066743 Chr.7: 36540318-36540318 on Build GRCh38
rs186405826 Chr.7: 36540279-36540279 on Build GRCh38
rs1001823985 Chr.7: 36540179-36540179 on Build GRCh38
rs917445706 Chr.7: 36540243-36540243 on Build GRCh38
rs953315148 Chr.7: 36540281-36540281 on Build GRCh38
rs1021276986 Chr.7: 36540137-36540137 on Build GRCh38
rs979512447 Chr.7: 36540196-36540196 on Build GRCh38
rs906020813 Chr.7: 36540165-36540165 on Build GRCh38
rs937780489 Chr.7: 36540205-36540205 on Build GRCh38
rs781647026 Chr.7: 36540277-36540277 on Build GRCh38
rs950184549 Chr.7: 36540245-36540245 on Build GRCh38
rs761825664 Chr.7: 36540198-36540198 on Build GRCh38
rs144072562 Chr.7: 36540132-36540142 on Build GRCh38
rs761689958 Chr.7: 36540329-36540329 on Build GRCh38
rs535988941 Chr.7: 36540175-36540175 on Build GRCh38
rs1038784119 Chr.7: 36540275-36540275 on Build GRCh38
rs370195853 Chr.7: 36540297-36540297 on Build GRCh38
rs1014635108 Chr.7: 36540206-36540206 on Build GRCh38

View Full Product Details