Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78589751-78589982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA5
Entrez Gene ID
1138
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
LNCR2
UniGene
Hs.1614
Gene Chromosome Location
Chr.15: 78565520-78595269 on Build GRCh38
Transcripts
NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5513115 c.425G>A p.R142H Chr.15: 78589816-78589816 on Build GRCh38
108281 c.533C>T p.P178L Chr.15: 78589924-78589924 on Build GRCh38
6891223 c.437C>T p.T146I Chr.15: 78589828-78589828 on Build GRCh38
3504316 c.421G>A p.G141R Chr.15: 78589812-78589812 on Build GRCh38
3504315 c.418G>A p.D140N Chr.15: 78589809-78589809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023953308 Chr.15: 78589951-78589951 on Build GRCh38
rs151206721 Chr.15: 78589929-78589929 on Build GRCh38
rs71528533 Chr.15: 78589792-78589792 on Build GRCh38
rs80087508 Chr.15: 78589891-78589891 on Build GRCh38
rs1043917444 Chr.15: 78589826-78589826 on Build GRCh38
rs779762437 Chr.15: 78589947-78589947 on Build GRCh38
rs754212902 Chr.15: 78589895-78589895 on Build GRCh38
rs763840987 Chr.15: 78589900-78589900 on Build GRCh38
rs200503890 Chr.15: 78589888-78589888 on Build GRCh38
rs762012646 Chr.15: 78589873-78589873 on Build GRCh38
rs200217616 Chr.15: 78589916-78589916 on Build GRCh38
rs774605972 Chr.15: 78589860-78589860 on Build GRCh38
rs749486168 Chr.15: 78589845-78589845 on Build GRCh38
rs375192419 Chr.15: 78589822-78589822 on Build GRCh38
rs373403458 Chr.15: 78589794-78589794 on Build GRCh38
rs751166834 Chr.15: 78589902-78589902 on Build GRCh38
rs199714438 Chr.15: 78589814-78589814 on Build GRCh38
rs781155351 Chr.15: 78589839-78589839 on Build GRCh38
rs375864160 Chr.15: 78589784-78589784 on Build GRCh38
rs55863434 Chr.15: 78589879-78589879 on Build GRCh38
rs780615731 Chr.15: 78589915-78589915 on Build GRCh38
rs937109017 Chr.15: 78589913-78589914 on Build GRCh38
rs77006957 Chr.15: 78589865-78589865 on Build GRCh38
rs756066200 Chr.15: 78589945-78589945 on Build GRCh38
rs949729557 Chr.15: 78589851-78589851 on Build GRCh38
rs200320233 Chr.15: 78589816-78589816 on Build GRCh38
rs199550371 Chr.15: 78589840-78589840 on Build GRCh38
rs773716974 Chr.15: 78589849-78589849 on Build GRCh38
rs574463276 Chr.15: 78589835-78589835 on Build GRCh38
rs370096844 Chr.15: 78589903-78589903 on Build GRCh38
rs111795555 Chr.15: 78589844-78589844 on Build GRCh38
rs151059425 Chr.15: 78589924-78589924 on Build GRCh38
rs773866112 Chr.15: 78589880-78589880 on Build GRCh38
rs201805555 Chr.15: 78589847-78589847 on Build GRCh38
rs576986903 Chr.15: 78589788-78589788 on Build GRCh38
rs201563436 Chr.15: 78589834-78589834 on Build GRCh38
rs765553764 Chr.15: 78589815-78589815 on Build GRCh38
rs765515682 Chr.15: 78589781-78589785 on Build GRCh38
rs71528534 Chr.15: 78589919-78589919 on Build GRCh38
rs762557487 Chr.15: 78589898-78589898 on Build GRCh38
rs200976675 Chr.15: 78589841-78589841 on Build GRCh38

View Full Product Details