Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150710970-150711241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN4
Entrez Gene ID
51164
Gene Name
dynactin subunit 4
Gene Aliases
DYN4, P62
UniGene
Hs.675564
Gene Chromosome Location
Chr.5: 150708747-150759095 on Build GRCh38
Transcripts
NM_001135643.1 NM_001135644.1 NM_016221.3 XM_005268457.1 XM_011537644.1 XM_011537645.1 XM_011537646.1 XM_011537647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4944195 c.1333A>T p.I445F Chr.5: 150711199-150711199 on Build GRCh38
4592564 c.1313G>A p.S438N Chr.5: 150711219-150711219 on Build GRCh38
1064436 c.1298G>A p.R433H Chr.5: 150711234-150711234 on Build GRCh38
6641138 c.1291C>A p.P431T Chr.5: 150711241-150711241 on Build GRCh38
1064437 c.1297C>T p.R433C Chr.5: 150711235-150711235 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763778489 Chr.5: 150711203-150711203 on Build GRCh38
rs201920711 Chr.5: 150711112-150711112 on Build GRCh38
rs781053588 Chr.5: 150711109-150711109 on Build GRCh38
rs760310764 Chr.5: 150711108-150711108 on Build GRCh38
rs1044678602 Chr.5: 150711019-150711019 on Build GRCh38
rs755217094 Chr.5: 150711135-150711135 on Build GRCh38
rs1054663661 Chr.5: 150710991-150710991 on Build GRCh38
rs536764862 Chr.5: 150711054-150711054 on Build GRCh38
rs3733923 Chr.5: 150711219-150711219 on Build GRCh38
rs948976589 Chr.5: 150711006-150711006 on Build GRCh38
rs748231357 Chr.5: 150711157-150711157 on Build GRCh38
rs1048975367 Chr.5: 150711035-150711035 on Build GRCh38
rs374616347 Chr.5: 150711124-150711124 on Build GRCh38
rs150282795 Chr.5: 150711213-150711213 on Build GRCh38
rs756969916 Chr.5: 150711046-150711046 on Build GRCh38
rs904885562 Chr.5: 150711030-150711030 on Build GRCh38
rs776186410 Chr.5: 150711211-150711211 on Build GRCh38
rs767251447 Chr.5: 150711101-150711101 on Build GRCh38
rs186059638 Chr.5: 150711007-150711007 on Build GRCh38
rs914823207 Chr.5: 150711001-150711001 on Build GRCh38
rs888760441 Chr.5: 150711058-150711058 on Build GRCh38
rs758993228 Chr.5: 150711127-150711127 on Build GRCh38
rs764787717 Chr.5: 150711217-150711217 on Build GRCh38
rs559647104 Chr.5: 150711175-150711175 on Build GRCh38
rs769297008 Chr.5: 150711027-150711027 on Build GRCh38
rs781321136 Chr.5: 150711155-150711155 on Build GRCh38
rs1056604314 Chr.5: 150711097-150711097 on Build GRCh38
rs780505866 Chr.5: 150711133-150711133 on Build GRCh38
rs775262271 Chr.5: 150711173-150711173 on Build GRCh38
rs573201219 Chr.5: 150711160-150711160 on Build GRCh38
rs540217397 Chr.5: 150711161-150711161 on Build GRCh38
rs377589711 Chr.5: 150711098-150711098 on Build GRCh38
rs866303066 Chr.5: 150711181-150711181 on Build GRCh38
rs199962532 Chr.5: 150711107-150711107 on Build GRCh38
rs765807312 Chr.5: 150711119-150711119 on Build GRCh38
rs747380320 Chr.5: 150711134-150711134 on Build GRCh38
rs772003011 Chr.5: 150711167-150711167 on Build GRCh38

View Full Product Details