Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7889559-7889832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
UniGene
Hs.25601
Gene Chromosome Location
Chr.17: 7884764-7912757 on Build GRCh38
Transcripts
NM_001005271.2 NM_001005273.2 NM_005852.3 XM_005256427.4 XM_005256428.4 XM_005256429.4 XM_005256431.4 XM_006721423.3 XM_006721424.3 XM_006721428.3 XM_017024061.1 XM_017024062.1 XM_017024063.1 XM_017024064.1 XM_017024065.1 XM_017024066.1 XM_017024067.1 XM_017024068.1 XM_017024069.1 XM_017024070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7179252 c.144G>C p.K48N Chr.17: 7889707-7889707 on Build GRCh38
4823217 c.115C>G p.R39G Chr.17: 7889678-7889678 on Build GRCh38
2802857 c.170_172delAGA p.K58delK Chr.17: 7889733-7889735 on Build GRCh38
985419 c.107A>G p.D36G Chr.17: 7889670-7889670 on Build GRCh38
5651050 c.155G>A p.R52Q Chr.17: 7889718-7889718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781050291 Chr.17: 7889787-7889787 on Build GRCh38
rs770986938 Chr.17: 7889660-7889660 on Build GRCh38
rs775748493 Chr.17: 7889696-7889696 on Build GRCh38
rs944874593 Chr.17: 7889695-7889695 on Build GRCh38
rs772231915 Chr.17: 7889689-7889689 on Build GRCh38
rs114834229 Chr.17: 7889590-7889590 on Build GRCh38
rs751355886 Chr.17: 7889776-7889776 on Build GRCh38
rs761069303 Chr.17: 7889712-7889712 on Build GRCh38
rs773838784 Chr.17: 7889669-7889669 on Build GRCh38
rs369508268 Chr.17: 7889797-7889797 on Build GRCh38
rs550038952 Chr.17: 7889765-7889765 on Build GRCh38
rs759766418 Chr.17: 7889675-7889675 on Build GRCh38
rs777179750 Chr.17: 7889733-7889733 on Build GRCh38
rs761220038 Chr.17: 7889781-7889781 on Build GRCh38
rs764553761 Chr.17: 7889730-7889730 on Build GRCh38
rs977948154 Chr.17: 7889737-7889737 on Build GRCh38
rs768510465 Chr.17: 7889648-7889648 on Build GRCh38
rs781153088 Chr.17: 7889649-7889649 on Build GRCh38
rs935878438 Chr.17: 7889634-7889634 on Build GRCh38
rs986310178 Chr.17: 7889616-7889616 on Build GRCh38
rs901914964 Chr.17: 7889783-7889783 on Build GRCh38
rs754780717 Chr.17: 7889785-7889785 on Build GRCh38
rs746773769 Chr.17: 7889629-7889629 on Build GRCh38
rs761473307 Chr.17: 7889758-7889758 on Build GRCh38
rs748114581 Chr.17: 7889788-7889788 on Build GRCh38
rs749377815 Chr.17: 7889654-7889654 on Build GRCh38
rs758150597 Chr.17: 7889774-7889774 on Build GRCh38
rs764841082 Chr.17: 7889760-7889760 on Build GRCh38
rs772319972 Chr.17: 7889810-7889810 on Build GRCh38
rs774624950 Chr.17: 7889672-7889672 on Build GRCh38
rs927199231 Chr.17: 7889631-7889631 on Build GRCh38
rs766019097 Chr.17: 7889775-7889775 on Build GRCh38
rs962409508 Chr.17: 7889582-7889582 on Build GRCh38
rs953572169 Chr.17: 7889609-7889609 on Build GRCh38
rs780011049 Chr.17: 7889622-7889622 on Build GRCh38
rs755986097 Chr.17: 7889796-7889796 on Build GRCh38
rs745908039 Chr.17: 7889805-7889805 on Build GRCh38
rs979731956 Chr.17: 7889597-7889597 on Build GRCh38
rs1054303002 Chr.17: 7889666-7889666 on Build GRCh38

View Full Product Details