Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14844424-14844691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs184385334 Chr.X: 14844493-14844493 on Build GRCh38
rs753885194 Chr.X: 14844642-14844642 on Build GRCh38
rs193205907 Chr.X: 14844463-14844463 on Build GRCh38
rs748424973 Chr.X: 14844462-14844465 on Build GRCh38
rs760863919 Chr.X: 14844597-14844597 on Build GRCh38
rs769508299 Chr.X: 14844568-14844568 on Build GRCh38
rs773176825 Chr.X: 14844575-14844575 on Build GRCh38
rs143850231 Chr.X: 14844517-14844517 on Build GRCh38
rs931259538 Chr.X: 14844618-14844618 on Build GRCh38
rs147255018 Chr.X: 14844578-14844578 on Build GRCh38
rs138268044 Chr.X: 14844511-14844511 on Build GRCh38
rs768048579 Chr.X: 14844538-14844538 on Build GRCh38
rs757409736 Chr.X: 14844464-14844464 on Build GRCh38
rs758926056 Chr.X: 14844509-14844509 on Build GRCh38
rs746890051 Chr.X: 14844566-14844566 on Build GRCh38
rs746571415 Chr.X: 14844513-14844513 on Build GRCh38
rs764091042 Chr.X: 14844615-14844615 on Build GRCh38
rs779253842 Chr.X: 14844480-14844480 on Build GRCh38
rs771044033 Chr.X: 14844584-14844584 on Build GRCh38
rs776403333 Chr.X: 14844542-14844542 on Build GRCh38
rs377268147 Chr.X: 14844653-14844653 on Build GRCh38
rs963760505 Chr.X: 14844443-14844443 on Build GRCh38
rs187737855 Chr.X: 14844596-14844596 on Build GRCh38
rs373141774 Chr.X: 14844522-14844522 on Build GRCh38
rs764440863 Chr.X: 14844460-14844460 on Build GRCh38
rs540569670 Chr.X: 14844577-14844577 on Build GRCh38
rs745346114 Chr.X: 14844668-14844668 on Build GRCh38
rs922251995 Chr.X: 14844474-14844474 on Build GRCh38
rs774837241 Chr.X: 14844459-14844468 on Build GRCh38
rs1039910199 Chr.X: 14844467-14844467 on Build GRCh38
rs868748586 Chr.X: 14844528-14844528 on Build GRCh38

View Full Product Details