Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83469176-83469449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG2
Entrez Gene ID
1740
Gene Name
discs large MAGUK scaffold protein 2
Gene Aliases
PPP1R58, PSD-93, PSD93, chapsyn-110
UniGene
Hs.367656
Gene Chromosome Location
Chr.11: 83455009-85628534 on Build GRCh38
Transcripts
NM_001142699.1 NM_001142700.1 NM_001142702.1 NM_001206769.1 NM_001300983.1 NM_001364.3 XM_005273810.4 XM_005273811.4 XM_011544778.2 XM_011544780.2 XM_011544782.2 XM_011544788.2 XM_017017254.1 XM_017017255.1 XM_017017256.1 XM_017017257.1 XM_017017258.1 XM_017017259.1 XM_017017260.1 XM_017017261.1 XM_017017262.1 XM_017017263.1 XM_017017264.1 XM_017017265.1 XM_017017266.1 XM_017017267.1 XM_017017268.1 XM_017017269.1 XM_017017270.1 XM_017017271.1 XM_017017272.1 XM_017017273.1 XM_017017274.1 XM_017017275.1 XM_017017276.1 XM_017017277.1 XM_017017278.1 XM_017017279.1 XM_017017280.1 XM_017017281.1 XM_017017282.1 XM_017017283.1 XM_017017284.1 XM_017017285.1 XM_017017286.1 XM_017017287.1 XM_017017288.1 XM_017017289.1 XM_017017290.1 XM_017017291.1 XM_017017292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5922884 c.1843G>A p.E615K Chr.11: 83469299-83469299 on Build GRCh38
3453964 c.2143C>T p.P715S Chr.11: 83469362-83469362 on Build GRCh38
6644680 c.2358C>T p.A786A Chr.11: 83469264-83469264 on Build GRCh38
6644681 c.2241C>T p.A747A Chr.11: 83469264-83469264 on Build GRCh38
1250171 c.2267G>T p.G756V Chr.11: 83469238-83469238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561760578 Chr.11: 83469243-83469243 on Build GRCh38
rs372440867 Chr.11: 83469285-83469285 on Build GRCh38
rs757300559 Chr.11: 83469416-83469416 on Build GRCh38
rs778065446 Chr.11: 83469413-83469413 on Build GRCh38
rs754190895 Chr.11: 83469366-83469366 on Build GRCh38
rs745502605 Chr.11: 83469273-83469273 on Build GRCh38
rs148801918 Chr.11: 83469372-83469372 on Build GRCh38
rs188274342 Chr.11: 83469404-83469404 on Build GRCh38
rs769362871 Chr.11: 83469277-83469277 on Build GRCh38
rs746732527 Chr.11: 83469247-83469247 on Build GRCh38
rs762467320 Chr.11: 83469308-83469308 on Build GRCh38
rs765399795 Chr.11: 83469400-83469400 on Build GRCh38
rs182838471 Chr.11: 83469264-83469264 on Build GRCh38
rs761225895 Chr.11: 83469334-83469334 on Build GRCh38
rs878923003 Chr.11: 83469407-83469407 on Build GRCh38
rs752657754 Chr.11: 83469384-83469385 on Build GRCh38
rs552118723 Chr.11: 83469384-83469384 on Build GRCh38
rs550649657 Chr.11: 83469341-83469341 on Build GRCh38
rs781001934 Chr.11: 83469419-83469419 on Build GRCh38
rs773707986 Chr.11: 83469332-83469332 on Build GRCh38
rs528157226 Chr.11: 83469205-83469205 on Build GRCh38
rs374845450 Chr.11: 83469278-83469278 on Build GRCh38
rs767948529 Chr.11: 83469328-83469328 on Build GRCh38
rs756046718 Chr.11: 83469394-83469399 on Build GRCh38
rs201804609 Chr.11: 83469225-83469225 on Build GRCh38
rs770843754 Chr.11: 83469255-83469255 on Build GRCh38
rs972171171 Chr.11: 83469392-83469392 on Build GRCh38
rs562758905 Chr.11: 83469348-83469348 on Build GRCh38
rs866533158 Chr.11: 83469402-83469402 on Build GRCh38

View Full Product Details