Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22593337-22593576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA8
Entrez Gene ID
2046
Gene Name
EPH receptor A8
Gene Aliases
EEK, EK3, HEK3
UniGene
Hs.283613
Gene Chromosome Location
Chr.1: 22563508-22603594 on Build GRCh38
Transcripts
XM_011540969.1 XM_011540970.1 XM_011540972.1 XM_011540973.1 XM_011540975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4751181 c.1333G>A p.V445M Chr.1: 22593343-22593343 on Build GRCh38
5019394 c.1401C>T p.N467N Chr.1: 22593411-22593411 on Build GRCh38
3484896 c.1467C>T p.T489T Chr.1: 22593550-22593550 on Build GRCh38
905219 c.1348C>T p.R450W Chr.1: 22593358-22593358 on Build GRCh38
4525853 c.1359G>T p.Q453H Chr.1: 22593369-22593369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771968074 Chr.1: 22593410-22593410 on Build GRCh38
rs374046323 Chr.1: 22593376-22593376 on Build GRCh38
rs768558919 Chr.1: 22593491-22593491 on Build GRCh38
rs757853715 Chr.1: 22593482-22593482 on Build GRCh38
rs780831697 Chr.1: 22593494-22593494 on Build GRCh38
rs749965171 Chr.1: 22593479-22593479 on Build GRCh38
rs761966259 Chr.1: 22593393-22593393 on Build GRCh38
rs758958503 Chr.1: 22593397-22593397 on Build GRCh38
rs199652427 Chr.1: 22593467-22593467 on Build GRCh38
rs755252711 Chr.1: 22593400-22593400 on Build GRCh38
rs77608596 Chr.1: 22593395-22593395 on Build GRCh38
rs778380990 Chr.1: 22593407-22593407 on Build GRCh38
rs757908930 Chr.1: 22593521-22593521 on Build GRCh38
rs756827993 Chr.1: 22593406-22593406 on Build GRCh38
rs781079148 Chr.1: 22593539-22593539 on Build GRCh38
rs371652847 Chr.1: 22593517-22593517 on Build GRCh38
rs777499719 Chr.1: 22593401-22593401 on Build GRCh38
rs779424856 Chr.1: 22593534-22593534 on Build GRCh38
rs563025239 Chr.1: 22593549-22593549 on Build GRCh38
rs765337661 Chr.1: 22593394-22593394 on Build GRCh38
rs199567884 Chr.1: 22593536-22593536 on Build GRCh38
rs770052567 Chr.1: 22593455-22593455 on Build GRCh38
rs968404464 Chr.1: 22593420-22593420 on Build GRCh38
rs932518395 Chr.1: 22593412-22593412 on Build GRCh38
rs867788111 Chr.1: 22593385-22593385 on Build GRCh38
rs572355548 Chr.1: 22593499-22593499 on Build GRCh38
rs209696 Chr.1: 22593411-22593411 on Build GRCh38
rs372181873 Chr.1: 22593466-22593466 on Build GRCh38
rs142120877 Chr.1: 22593540-22593540 on Build GRCh38
rs369922049 Chr.1: 22593375-22593375 on Build GRCh38
rs374661974 Chr.1: 22593359-22593359 on Build GRCh38
rs767772053 Chr.1: 22593465-22593465 on Build GRCh38
rs759953301 Chr.1: 22593461-22593461 on Build GRCh38
rs778436054 Chr.1: 22593469-22593469 on Build GRCh38
rs376945590 Chr.1: 22593408-22593408 on Build GRCh38
rs770105897 Chr.1: 22593498-22593498 on Build GRCh38
rs200728689 Chr.1: 22593511-22593511 on Build GRCh38
rs199607831 Chr.1: 22593457-22593457 on Build GRCh38
rs201254867 Chr.1: 22593493-22593493 on Build GRCh38
rs760846218 Chr.1: 22593363-22593363 on Build GRCh38
rs751955835 Chr.1: 22593458-22593458 on Build GRCh38
rs761932406 Chr.1: 22593445-22593445 on Build GRCh38
rs766896165 Chr.1: 22593399-22593399 on Build GRCh38
rs754845150 Chr.1: 22593538-22593538 on Build GRCh38
rs376592875 Chr.1: 22593502-22593502 on Build GRCh38
rs760989055 Chr.1: 22593515-22593515 on Build GRCh38
rs369590570 Chr.1: 22593510-22593510 on Build GRCh38
rs149335835 Chr.1: 22593555-22593555 on Build GRCh38
rs746841294 Chr.1: 22593415-22593415 on Build GRCh38
rs535468226 Chr.1: 22593444-22593444 on Build GRCh38
rs375134443 Chr.1: 22593516-22593516 on Build GRCh38
rs748829132 Chr.1: 22593405-22593405 on Build GRCh38
rs374051191 Chr.1: 22593485-22593485 on Build GRCh38
rs751569232 Chr.1: 22593535-22593535 on Build GRCh38
rs775244487 Chr.1: 22593362-22593362 on Build GRCh38
rs895380846 Chr.1: 22593520-22593520 on Build GRCh38
rs768277316 Chr.1: 22593429-22593429 on Build GRCh38
rs115749606 Chr.1: 22593512-22593512 on Build GRCh38
rs746854511 Chr.1: 22593490-22593490 on Build GRCh38

View Full Product Details