Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7075113-7075346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC10A
Entrez Gene ID
10462
Gene Name
C-type lectin domain family 10 member A
Gene Aliases
CD301, CLECSF13, CLECSF14, HML, HML2, MGL
UniGene
Hs.54403
Gene Chromosome Location
Chr.17: 7074537-7080635 on Build GRCh38
Transcripts
NM_006344.2 NM_182906.2 XM_005256412.1 XM_011523613.1 XM_011523614.2 XM_011523615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2153435 c.855C>T p.F285F Chr.17: 7075150-7075150 on Build GRCh38
5782965 c.869G>T p.R290M Chr.17: 7075136-7075136 on Build GRCh38
1743027 c.823G>A p.G275R Chr.17: 7075182-7075182 on Build GRCh38
4672706 c.838G>A p.E280K Chr.17: 7075167-7075167 on Build GRCh38
6630077 c.827T>C p.L276P Chr.17: 7075178-7075178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759641268 Chr.17: 7075158-7075159 on Build GRCh38
rs778981235 Chr.17: 7075326-7075326 on Build GRCh38
rs776162818 Chr.17: 7075223-7075223 on Build GRCh38
rs751782878 Chr.17: 7075262-7075262 on Build GRCh38
rs780248039 Chr.17: 7075150-7075150 on Build GRCh38
rs200496384 Chr.17: 7075197-7075197 on Build GRCh38
rs377248841 Chr.17: 7075317-7075317 on Build GRCh38
rs140942315 Chr.17: 7075196-7075196 on Build GRCh38
rs146480775 Chr.17: 7075185-7075185 on Build GRCh38
rs372729049 Chr.17: 7075141-7075141 on Build GRCh38
rs375550026 Chr.17: 7075164-7075164 on Build GRCh38
rs906818959 Chr.17: 7075304-7075304 on Build GRCh38
rs115347328 Chr.17: 7075182-7075182 on Build GRCh38
rs149746467 Chr.17: 7075139-7075139 on Build GRCh38
rs768084979 Chr.17: 7075211-7075211 on Build GRCh38
rs145596692 Chr.17: 7075167-7075167 on Build GRCh38
rs767516359 Chr.17: 7075319-7075319 on Build GRCh38
rs142642957 Chr.17: 7075183-7075183 on Build GRCh38
rs752798672 Chr.17: 7075323-7075323 on Build GRCh38
rs766668841 Chr.17: 7075254-7075254 on Build GRCh38
rs749215238 Chr.17: 7075168-7075168 on Build GRCh38
rs762279650 Chr.17: 7075224-7075224 on Build GRCh38
rs773628634 Chr.17: 7075227-7075227 on Build GRCh38
rs991113331 Chr.17: 7075140-7075140 on Build GRCh38
rs948084991 Chr.17: 7075213-7075213 on Build GRCh38
rs570753508 Chr.17: 7075277-7075277 on Build GRCh38
rs765740164 Chr.17: 7075225-7075225 on Build GRCh38
rs368116399 Chr.17: 7075324-7075324 on Build GRCh38
rs371677191 Chr.17: 7075291-7075291 on Build GRCh38
rs112204401 Chr.17: 7075198-7075198 on Build GRCh38
rs556438794 Chr.17: 7075327-7075327 on Build GRCh38
rs763287936 Chr.17: 7075246-7075246 on Build GRCh38
rs751692946 Chr.17: 7075158-7075158 on Build GRCh38
rs552400469 Chr.17: 7075133-7075133 on Build GRCh38

View Full Product Details