Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 28995739-28996012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPVL
Entrez Gene ID
54504
Gene Name
carboxypeptidase, vitellogenic like
Gene Aliases
HVLP
UniGene
Hs.233389
Gene Chromosome Location
Chr.7: 28995630-29146537 on Build GRCh38
Transcripts
NM_019029.2 NM_031311.3 XM_011515437.1 XM_017012366.1 NR_038965.1

Gene Symbol
LOC100506497
Entrez Gene ID
100506497
Gene Name
uncharacterized LOC100506497
Gene Aliases
-
UniGene
Hs.586328
Gene Chromosome Location
Chr.7: 28979967-29013369 on Build GRCh38
Transcripts
NM_019029.2 NM_031311.3 XM_011515437.1 XM_017012366.1 NR_038965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4638632 c.1331G>C p.R444P Chr.7: 28995872-28995872 on Build GRCh38
232090 c.1364C>T p.P455L Chr.7: 28995839-28995839 on Build GRCh38
6634995 c.1383G>T p.M461I Chr.7: 28995820-28995820 on Build GRCh38
214097 c.1429T>A p.*477K Chr.7: 28995774-28995774 on Build GRCh38
1088940 c.1391G>A p.R464Q Chr.7: 28995812-28995812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779911216 Chr.7: 28995847-28995847 on Build GRCh38
rs2302028 Chr.7: 28995913-28995913 on Build GRCh38
rs147771477 Chr.7: 28995812-28995812 on Build GRCh38
rs749093747 Chr.7: 28995849-28995849 on Build GRCh38
rs748029684 Chr.7: 28995861-28995861 on Build GRCh38
rs201351381 Chr.7: 28995873-28995873 on Build GRCh38
rs895724955 Chr.7: 28995771-28995771 on Build GRCh38
rs757457600 Chr.7: 28995809-28995809 on Build GRCh38
rs375370289 Chr.7: 28995835-28995835 on Build GRCh38
rs1038595717 Chr.7: 28995852-28995852 on Build GRCh38
rs961112662 Chr.7: 28995882-28995882 on Build GRCh38
rs373399772 Chr.7: 28995779-28995779 on Build GRCh38
rs943518947 Chr.7: 28995942-28995942 on Build GRCh38
rs200592788 Chr.7: 28995908-28995908 on Build GRCh38
rs766008084 Chr.7: 28995879-28995879 on Build GRCh38
rs10228797 Chr.7: 28995938-28995938 on Build GRCh38
rs776344521 Chr.7: 28995895-28995895 on Build GRCh38
rs762365980 Chr.7: 28995907-28995907 on Build GRCh38
rs746123081 Chr.7: 28995816-28995816 on Build GRCh38
rs369048385 Chr.7: 28995925-28995925 on Build GRCh38
rs763264897 Chr.7: 28995896-28995896 on Build GRCh38
rs978892209 Chr.7: 28995975-28995975 on Build GRCh38
rs764573965 Chr.7: 28995793-28995793 on Build GRCh38
rs1022608966 Chr.7: 28995958-28995958 on Build GRCh38
rs572882434 Chr.7: 28995981-28995981 on Build GRCh38
rs751067717 Chr.7: 28995886-28995891 on Build GRCh38
rs769170811 Chr.7: 28995893-28995898 on Build GRCh38
rs151217521 Chr.7: 28995781-28995781 on Build GRCh38
rs751767950 Chr.7: 28995916-28995916 on Build GRCh38
rs1034734650 Chr.7: 28995930-28995930 on Build GRCh38
rs369717141 Chr.7: 28995906-28995906 on Build GRCh38
rs771602383 Chr.7: 28995872-28995872 on Build GRCh38
rs774205940 Chr.7: 28995857-28995857 on Build GRCh38
rs764493990 Chr.7: 28995900-28995900 on Build GRCh38
rs760285647 Chr.7: 28995778-28995778 on Build GRCh38
rs758780191 Chr.7: 28995788-28995788 on Build GRCh38
rs768468978 Chr.7: 28995854-28995854 on Build GRCh38
rs201613280 Chr.7: 28995780-28995780 on Build GRCh38

View Full Product Details