Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43360793-43361048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
UniGene
Hs.164453
Gene Chromosome Location
Chr.7: 43112580-43566001 on Build GRCh38
Transcripts
NM_001287059.1 NM_015052.4 XM_005249665.3 XM_006715670.3 XM_006715671.3 XM_006715673.3 XM_011515220.2 XM_011515222.2 XM_011515223.2 XM_011515224.2 XM_011515225.2 XM_017011882.1 XM_017011883.1 XM_017011884.1 XM_017011885.1 XM_017011886.1 XM_017011887.1 XM_017011888.1 XM_017011889.1 XM_017011890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
746879 c.533C>A p.T178K Chr.7: 43360958-43360958 on Build GRCh38
5674606 c.521C>G p.T174S Chr.7: 43360946-43360946 on Build GRCh38
4424568 c.463G>A p.E155K Chr.7: 43360888-43360888 on Build GRCh38
95938 c.507C>A p.A169A Chr.7: 43360932-43360932 on Build GRCh38
4644800 c.550G>T p.A184S Chr.7: 43360975-43360975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926464487 Chr.7: 43360865-43360865 on Build GRCh38
rs369654096 Chr.7: 43360971-43360971 on Build GRCh38
rs773219733 Chr.7: 43360925-43360925 on Build GRCh38
rs915147710 Chr.7: 43360848-43360848 on Build GRCh38
rs946634070 Chr.7: 43360851-43360851 on Build GRCh38
rs768784411 Chr.7: 43361016-43361016 on Build GRCh38
rs781620445 Chr.7: 43360846-43360846 on Build GRCh38
rs564813951 Chr.7: 43360817-43360817 on Build GRCh38
rs749449401 Chr.7: 43360877-43360877 on Build GRCh38
rs573464359 Chr.7: 43360832-43360832 on Build GRCh38
rs375316956 Chr.7: 43360921-43360921 on Build GRCh38
rs760588791 Chr.7: 43360926-43360926 on Build GRCh38
rs756729920 Chr.7: 43360840-43360841 on Build GRCh38
rs764819634 Chr.7: 43360968-43360968 on Build GRCh38
rs750881356 Chr.7: 43360979-43360979 on Build GRCh38
rs746234676 Chr.7: 43360855-43360855 on Build GRCh38
rs995260430 Chr.7: 43360860-43360860 on Build GRCh38
rs76459581 Chr.7: 43360997-43360997 on Build GRCh38
rs142171339 Chr.7: 43360998-43360998 on Build GRCh38
rs753563877 Chr.7: 43360949-43360949 on Build GRCh38
rs759139966 Chr.7: 43360966-43360966 on Build GRCh38
rs774605928 Chr.7: 43360913-43360913 on Build GRCh38
rs768822348 Chr.7: 43360894-43360894 on Build GRCh38
rs757780423 Chr.7: 43360838-43360838 on Build GRCh38
rs775838268 Chr.7: 43360873-43360873 on Build GRCh38
rs540865392 Chr.7: 43360972-43360972 on Build GRCh38
rs766078063 Chr.7: 43360937-43360937 on Build GRCh38
rs770049408 Chr.7: 43360866-43360866 on Build GRCh38
rs780873522 Chr.7: 43361001-43361002 on Build GRCh38
rs771280824 Chr.7: 43360959-43360959 on Build GRCh38
rs781713887 Chr.7: 43360975-43360975 on Build GRCh38
rs991570412 Chr.7: 43360994-43360994 on Build GRCh38
rs369959223 Chr.7: 43361005-43361005 on Build GRCh38
rs372011835 Chr.7: 43360917-43360917 on Build GRCh38

View Full Product Details