Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50067935-50068196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA3
Entrez Gene ID
3675
Gene Name
integrin subunit alpha 3
Gene Aliases
CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a
UniGene
Hs.265829
Gene Chromosome Location
Chr.17: 50055968-50090485 on Build GRCh38
Transcripts
NM_002204.3 XM_005257308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4493589 c.417C>T p.V139V Chr.17: 50068058-50068058 on Build GRCh38
1384135 c.467G>T p.R156L Chr.17: 50068108-50068108 on Build GRCh38
1588963 c.496G>A p.G166S Chr.17: 50068137-50068137 on Build GRCh38
4794081 c.531G>A p.W177* Chr.17: 50068172-50068172 on Build GRCh38
4067537 c.493C>T p.R165* Chr.17: 50068134-50068134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755023428 Chr.17: 50068125-50068125 on Build GRCh38
rs763799707 Chr.17: 50068069-50068069 on Build GRCh38
rs978968835 Chr.17: 50068021-50068021 on Build GRCh38
rs748413546 Chr.17: 50068029-50068029 on Build GRCh38
rs534708438 Chr.17: 50067995-50067995 on Build GRCh38
rs755144136 Chr.17: 50068015-50068015 on Build GRCh38
rs993111985 Chr.17: 50068002-50068002 on Build GRCh38
rs142114722 Chr.17: 50067987-50067987 on Build GRCh38
rs557579280 Chr.17: 50068110-50068110 on Build GRCh38
rs761605539 Chr.17: 50068102-50068102 on Build GRCh38
rs773509079 Chr.17: 50068043-50068043 on Build GRCh38
rs771415485 Chr.17: 50068161-50068161 on Build GRCh38
rs750191749 Chr.17: 50068108-50068108 on Build GRCh38
rs749625688 Chr.17: 50068141-50068141 on Build GRCh38
rs946000001 Chr.17: 50068083-50068083 on Build GRCh38
rs751619579 Chr.17: 50068124-50068124 on Build GRCh38
rs746374117 Chr.17: 50068042-50068043 on Build GRCh38
rs766249771 Chr.17: 50068111-50068111 on Build GRCh38
rs536809708 Chr.17: 50068131-50068131 on Build GRCh38
rs750350999 Chr.17: 50068005-50068005 on Build GRCh38
rs368137808 Chr.17: 50068068-50068068 on Build GRCh38
rs747073365 Chr.17: 50068013-50068013 on Build GRCh38
rs770124636 Chr.17: 50068037-50068037 on Build GRCh38
rs567819625 Chr.17: 50068063-50068063 on Build GRCh38
rs938402107 Chr.17: 50067990-50067990 on Build GRCh38
rs756398910 Chr.17: 50068134-50068134 on Build GRCh38
rs536104693 Chr.17: 50068082-50068082 on Build GRCh38
rs749795382 Chr.17: 50068044-50068044 on Build GRCh38
rs771343383 Chr.17: 50068048-50068048 on Build GRCh38
rs780116501 Chr.17: 50068009-50068009 on Build GRCh38
rs776392266 Chr.17: 50068095-50068095 on Build GRCh38
rs781302203 Chr.17: 50068130-50068130 on Build GRCh38
rs1020543830 Chr.17: 50068006-50068006 on Build GRCh38
rs773908262 Chr.17: 50067960-50067960 on Build GRCh38
rs180868125 Chr.17: 50068019-50068019 on Build GRCh38
rs761431632 Chr.17: 50068008-50068008 on Build GRCh38
rs778130201 Chr.17: 50068135-50068135 on Build GRCh38
rs369557937 Chr.17: 50068107-50068107 on Build GRCh38

View Full Product Details