Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99685691-99685964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99659506-99711245 on Build GRCh38
Transcripts
NM_020354.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
913775 c.645G>A p.K215K Chr.10: 99685888-99685888 on Build GRCh38
2057408 c.574C>A p.L192I Chr.10: 99685817-99685817 on Build GRCh38
7180078 c.609C>G p.L203L Chr.10: 99685852-99685852 on Build GRCh38
2057409 c.597G>T p.E199D Chr.10: 99685840-99685840 on Build GRCh38
6654348 c.637T>G p.S213A Chr.10: 99685880-99685880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777299636 Chr.10: 99685785-99685785 on Build GRCh38
rs200454165 Chr.10: 99685850-99685850 on Build GRCh38
rs370666357 Chr.10: 99685805-99685805 on Build GRCh38
rs989530425 Chr.10: 99685727-99685727 on Build GRCh38
rs750886155 Chr.10: 99685898-99685898 on Build GRCh38
rs769675684 Chr.10: 99685837-99685837 on Build GRCh38
rs768883678 Chr.10: 99685852-99685852 on Build GRCh38
rs990005389 Chr.10: 99685754-99685754 on Build GRCh38
rs747670371 Chr.10: 99685781-99685781 on Build GRCh38
rs887282095 Chr.10: 99685921-99685921 on Build GRCh38
rs1021057562 Chr.10: 99685770-99685770 on Build GRCh38
rs915200731 Chr.10: 99685744-99685744 on Build GRCh38
rs756734808 Chr.10: 99685874-99685874 on Build GRCh38
rs372263602 Chr.10: 99685912-99685912 on Build GRCh38
rs780949116 Chr.10: 99685938-99685938 on Build GRCh38
rs113578309 Chr.10: 99685810-99685810 on Build GRCh38
rs1047433727 Chr.10: 99685940-99685940 on Build GRCh38
rs769940879 Chr.10: 99685765-99685766 on Build GRCh38
rs777437077 Chr.10: 99685927-99685927 on Build GRCh38
rs757881619 Chr.10: 99685915-99685915 on Build GRCh38
rs775580618 Chr.10: 99685840-99685840 on Build GRCh38
rs767963453 Chr.10: 99685892-99685892 on Build GRCh38
rs928190238 Chr.10: 99685818-99685818 on Build GRCh38
rs368192875 Chr.10: 99685899-99685899 on Build GRCh38
rs201287793 Chr.10: 99685842-99685842 on Build GRCh38
rs940383574 Chr.10: 99685922-99685922 on Build GRCh38
rs928193032 Chr.10: 99685873-99685873 on Build GRCh38
rs956757216 Chr.10: 99685714-99685714 on Build GRCh38
rs770686860 Chr.10: 99685814-99685814 on Build GRCh38
rs3740078 Chr.10: 99685834-99685834 on Build GRCh38
rs756966654 Chr.10: 99685928-99685928 on Build GRCh38
rs778483857 Chr.10: 99685752-99685752 on Build GRCh38
rs991002328 Chr.10: 99685835-99685835 on Build GRCh38
rs758014656 Chr.10: 99685783-99685783 on Build GRCh38
rs775701898 Chr.10: 99685768-99685769 on Build GRCh38
rs199518762 Chr.10: 99685909-99685909 on Build GRCh38
rs376174841 Chr.10: 99685788-99685788 on Build GRCh38
rs776446713 Chr.10: 99685827-99685827 on Build GRCh38

View Full Product Details