Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 105233609-105233882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RAPL2
Entrez Gene ID
26280
Gene Name
interleukin 1 receptor accessory protein like 2
Gene Aliases
IL-1R9, IL1R9, IL1RAPL-2, TIGIRR-1
UniGene
Hs.675519
Gene Chromosome Location
Chr.X: 104566315-105769729 on Build GRCh38
Transcripts
NM_017416.1 XM_011530905.2 XM_017029403.1

SNP Detail

SNP ID Chromosome Location
rs1048715439 Chr.X: 105233769-105233769 on Build GRCh38
rs187161585 Chr.X: 105233784-105233784 on Build GRCh38
rs782200378 Chr.X: 105233696-105233697 on Build GRCh38
rs782449569 Chr.X: 105233851-105233851 on Build GRCh38
rs782232010 Chr.X: 105233797-105233797 on Build GRCh38
rs781835243 Chr.X: 105233765-105233765 on Build GRCh38
rs948184452 Chr.X: 105233805-105233805 on Build GRCh38
rs782475663 Chr.X: 105233645-105233645 on Build GRCh38
rs782760439 Chr.X: 105233698-105233698 on Build GRCh38
rs782635783 Chr.X: 105233650-105233650 on Build GRCh38
rs782804693 Chr.X: 105233775-105233775 on Build GRCh38
rs782709147 Chr.X: 105233827-105233827 on Build GRCh38
rs782066758 Chr.X: 105233798-105233798 on Build GRCh38
rs782651222 Chr.X: 105233789-105233789 on Build GRCh38
rs182129432 Chr.X: 105233781-105233781 on Build GRCh38
rs782563129 Chr.X: 105233858-105233858 on Build GRCh38
rs554371510 Chr.X: 105233669-105233669 on Build GRCh38
rs976729889 Chr.X: 105233682-105233682 on Build GRCh38

View Full Product Details