Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104392080-104392335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP58
Entrez Gene ID
159686
Gene Name
cilia and flagella associated protein 58
Gene Aliases
C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1
UniGene
Hs.253576
Gene Chromosome Location
Chr.10: 104339124-104455090 on Build GRCh38
Transcripts
NM_001008723.1 XM_011539377.2 XM_017015787.1 XM_017015788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7092555 c.1465A>C p.N489H Chr.10: 104392332-104392332 on Build GRCh38
683431 c.1462C>G p.Q488E Chr.10: 104392329-104392329 on Build GRCh38
1627303 c.1427A>G p.K476R Chr.10: 104392294-104392294 on Build GRCh38
1247471 c.1423delA p.I477fs*1 Chr.10: 104392290-104392290 on Build GRCh38
5359811 c.1407G>T p.Q469H Chr.10: 104392274-104392274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757444655 Chr.10: 104392257-104392257 on Build GRCh38
rs911275019 Chr.10: 104392166-104392166 on Build GRCh38
rs865850466 Chr.10: 104392247-104392247 on Build GRCh38
rs886742615 Chr.10: 104392124-104392124 on Build GRCh38
rs755743991 Chr.10: 104392198-104392198 on Build GRCh38
rs761844995 Chr.10: 104392290-104392290 on Build GRCh38
rs536262017 Chr.10: 104392167-104392167 on Build GRCh38
rs1045415891 Chr.10: 104392118-104392118 on Build GRCh38
rs779708447 Chr.10: 104392205-104392205 on Build GRCh38
rs369038905 Chr.10: 104392229-104392229 on Build GRCh38
rs552993304 Chr.10: 104392178-104392178 on Build GRCh38
rs778665302 Chr.10: 104392220-104392220 on Build GRCh38
rs112168082 Chr.10: 104392264-104392264 on Build GRCh38
rs753867814 Chr.10: 104392217-104392217 on Build GRCh38
rs771696882 Chr.10: 104392230-104392230 on Build GRCh38
rs56655226 Chr.10: 104392190-104392208 on Build GRCh38
rs777981562 Chr.10: 104392263-104392263 on Build GRCh38
rs201667625 Chr.10: 104392204-104392204 on Build GRCh38
rs920100049 Chr.10: 104392251-104392251 on Build GRCh38
rs1003967530 Chr.10: 104392153-104392153 on Build GRCh38
rs191921926 Chr.10: 104392140-104392140 on Build GRCh38
rs767297659 Chr.10: 104392289-104392290 on Build GRCh38
rs771062873 Chr.10: 104392288-104392288 on Build GRCh38
rs376065077 Chr.10: 104392289-104392289 on Build GRCh38

View Full Product Details