Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85401100-85401365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPG
Entrez Gene ID
822
Gene Name
capping actin protein, gelsolin like
Gene Aliases
AFCP, HEL-S-66, MCP
UniGene
Hs.516155
Gene Chromosome Location
Chr.2: 85394748-85418467 on Build GRCh38
Transcripts
NM_001256139.1 NM_001256140.1 NM_001320732.1 NM_001320733.1 NM_001320734.1 NM_001747.3 XM_011533122.1 XM_011533123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3391876 c.469T>C p.W157R Chr.2: 85401212-85401212 on Build GRCh38
3659996 c.514C>A p.Q172K Chr.2: 85401167-85401167 on Build GRCh38
6092490 c.430G>A p.G144R Chr.2: 85401251-85401251 on Build GRCh38
6619723 c.378C>T p.H126H Chr.2: 85401303-85401303 on Build GRCh38
3001690 c.498C>T p.F166F Chr.2: 85401183-85401183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778854503 Chr.2: 85401322-85401322 on Build GRCh38
rs910846230 Chr.2: 85401258-85401258 on Build GRCh38
rs1048521847 Chr.2: 85401250-85401250 on Build GRCh38
rs771158420 Chr.2: 85401123-85401123 on Build GRCh38
rs377002350 Chr.2: 85401217-85401217 on Build GRCh38
rs139719975 Chr.2: 85401265-85401265 on Build GRCh38
rs779823535 Chr.2: 85401182-85401182 on Build GRCh38
rs199752504 Chr.2: 85401135-85401135 on Build GRCh38
rs749881481 Chr.2: 85401173-85401173 on Build GRCh38
rs775560591 Chr.2: 85401136-85401136 on Build GRCh38
rs753552891 Chr.2: 85401310-85401310 on Build GRCh38
rs778524584 Chr.2: 85401191-85401191 on Build GRCh38
rs866235573 Chr.2: 85401311-85401311 on Build GRCh38
rs750929355 Chr.2: 85401266-85401266 on Build GRCh38
rs764330765 Chr.2: 85401157-85401157 on Build GRCh38
rs1035036141 Chr.2: 85401211-85401211 on Build GRCh38
rs896077843 Chr.2: 85401327-85401327 on Build GRCh38
rs769755908 Chr.2: 85401228-85401228 on Build GRCh38
rs200720697 Chr.2: 85401236-85401236 on Build GRCh38
rs776811032 Chr.2: 85401129-85401129 on Build GRCh38
rs202207205 Chr.2: 85401164-85401164 on Build GRCh38
rs142762000 Chr.2: 85401227-85401227 on Build GRCh38
rs769962023 Chr.2: 85401343-85401343 on Build GRCh38
rs1040571244 Chr.2: 85401293-85401293 on Build GRCh38
rs760273912 Chr.2: 85401273-85401273 on Build GRCh38
rs1035978746 Chr.2: 85401317-85401317 on Build GRCh38
rs777711420 Chr.2: 85401263-85401264 on Build GRCh38
rs752107988 Chr.2: 85401161-85401161 on Build GRCh38
rs776864518 Chr.2: 85401226-85401226 on Build GRCh38
rs553867141 Chr.2: 85401255-85401255 on Build GRCh38
rs1004868172 Chr.2: 85401267-85401267 on Build GRCh38
rs185763147 Chr.2: 85401224-85401224 on Build GRCh38
rs566095029 Chr.2: 85401320-85401320 on Build GRCh38
rs755570197 Chr.2: 85401174-85401174 on Build GRCh38
rs369940669 Chr.2: 85401335-85401335 on Build GRCh38
rs373837301 Chr.2: 85401139-85401139 on Build GRCh38
rs370122534 Chr.2: 85401284-85401284 on Build GRCh38
rs375419436 Chr.2: 85401274-85401274 on Build GRCh38
rs866509203 Chr.2: 85401305-85401305 on Build GRCh38
rs988262377 Chr.2: 85401221-85401221 on Build GRCh38
rs568415860 Chr.2: 85401188-85401188 on Build GRCh38
rs748861828 Chr.2: 85401187-85401187 on Build GRCh38
rs767022200 Chr.2: 85401171-85401171 on Build GRCh38
rs111495086 Chr.2: 85401235-85401235 on Build GRCh38
rs765785513 Chr.2: 85401303-85401303 on Build GRCh38
rs1039561537 Chr.2: 85401229-85401229 on Build GRCh38
rs752480799 Chr.2: 85401333-85401333 on Build GRCh38
rs539550972 Chr.2: 85401342-85401342 on Build GRCh38
rs867161565 Chr.2: 85401191-85401191 on Build GRCh38
rs775817650 Chr.2: 85401232-85401232 on Build GRCh38
rs913957735 Chr.2: 85401223-85401223 on Build GRCh38

View Full Product Details