Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37862234-37862477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5B
Entrez Gene ID
3633
Gene Name
inositol polyphosphate-5-phosphatase B
Gene Aliases
5PTase
UniGene
Hs.449942
Gene Chromosome Location
Chr.1: 37860697-37947094 on Build GRCh38
Transcripts
NM_001297434.1 NM_005540.2 XM_006710621.3 XM_011541392.2 XM_011541393.2 XM_017001207.1 XM_017001208.1 XM_017001209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4007640 c.2936A>C p.K979T Chr.1: 37862361-37862361 on Build GRCh38
7372281 c.2920G>C p.D974H Chr.1: 37862377-37862377 on Build GRCh38
7274628 c.2925G>A p.M975I Chr.1: 37862372-37862372 on Build GRCh38
2167642 c.2894G>A p.R965Q Chr.1: 37862403-37862403 on Build GRCh38
909070 c.2951T>G p.F984C Chr.1: 37862346-37862346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763066919 Chr.1: 37862321-37862321 on Build GRCh38
rs571764941 Chr.1: 37862386-37862386 on Build GRCh38
rs770754718 Chr.1: 37862357-37862359 on Build GRCh38
rs776262067 Chr.1: 37862410-37862410 on Build GRCh38
rs755928040 Chr.1: 37862425-37862425 on Build GRCh38
rs770997958 Chr.1: 37862435-37862435 on Build GRCh38
rs191075425 Chr.1: 37862376-37862376 on Build GRCh38
rs770035940 Chr.1: 37862318-37862318 on Build GRCh38
rs369917721 Chr.1: 37862301-37862301 on Build GRCh38
rs561088456 Chr.1: 37862358-37862358 on Build GRCh38
rs751575274 Chr.1: 37862336-37862336 on Build GRCh38
rs527541022 Chr.1: 37862277-37862277 on Build GRCh38
rs772988409 Chr.1: 37862265-37862266 on Build GRCh38
rs373776316 Chr.1: 37862389-37862389 on Build GRCh38
rs140670679 Chr.1: 37862264-37862265 on Build GRCh38
rs773701071 Chr.1: 37862281-37862281 on Build GRCh38
rs755827992 Chr.1: 37862265-37862265 on Build GRCh38
rs745882924 Chr.1: 37862285-37862285 on Build GRCh38
rs3843 Chr.1: 37862310-37862310 on Build GRCh38
rs980 Chr.1: 37862320-37862320 on Build GRCh38
rs187077116 Chr.1: 37862302-37862302 on Build GRCh38
rs370532382 Chr.1: 37862432-37862432 on Build GRCh38
rs375829441 Chr.1: 37862404-37862404 on Build GRCh38
rs756853976 Chr.1: 37862373-37862373 on Build GRCh38
rs374940636 Chr.1: 37862442-37862442 on Build GRCh38
rs529874480 Chr.1: 37862359-37862359 on Build GRCh38
rs200258085 Chr.1: 37862264-37862264 on Build GRCh38
rs980916507 Chr.1: 37862364-37862364 on Build GRCh38
rs779926757 Chr.1: 37862395-37862395 on Build GRCh38
rs746831813 Chr.1: 37862403-37862403 on Build GRCh38
rs370097490 Chr.1: 37862335-37862335 on Build GRCh38
rs748769917 Chr.1: 37862270-37862270 on Build GRCh38
rs768065842 Chr.1: 37862353-37862353 on Build GRCh38
rs774299052 Chr.1: 37862437-37862438 on Build GRCh38
rs549738060 Chr.1: 37862342-37862342 on Build GRCh38
rs777523054 Chr.1: 37862267-37862267 on Build GRCh38
rs753565968 Chr.1: 37862370-37862370 on Build GRCh38
rs543216949 Chr.1: 37862299-37862301 on Build GRCh38
rs766524381 Chr.1: 37862327-37862327 on Build GRCh38
rs745312224 Chr.1: 37862378-37862378 on Build GRCh38

View Full Product Details