Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48603595-48603834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83E
Entrez Gene ID
54854
Gene Name
family with sequence similarity 83 member E
Gene Aliases
-
UniGene
Hs.165803
Gene Chromosome Location
Chr.19: 48600600-48614841 on Build GRCh38
Transcripts
NM_017708.3 XM_005259019.1 XM_011527049.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4614848 c.889delC p.Q297fs*6 Chr.19: 48603781-48603781 on Build GRCh38
1132995 c.932C>T p.P311L Chr.19: 48603738-48603738 on Build GRCh38
3536571 c.878C>T p.P293L Chr.19: 48603792-48603792 on Build GRCh38
3536572 c.868C>T p.P290S Chr.19: 48603802-48603802 on Build GRCh38
1395068 c.869C>T p.P290L Chr.19: 48603801-48603801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762174581 Chr.19: 48603633-48603633 on Build GRCh38
rs747074642 Chr.19: 48603674-48603674 on Build GRCh38
rs773896990 Chr.19: 48603687-48603687 on Build GRCh38
rs752338055 Chr.19: 48603744-48603744 on Build GRCh38
rs751033285 Chr.19: 48603699-48603699 on Build GRCh38
rs138269435 Chr.19: 48603678-48603678 on Build GRCh38
rs536095926 Chr.19: 48603618-48603618 on Build GRCh38
rs776858937 Chr.19: 48603680-48603680 on Build GRCh38
rs773971919 Chr.19: 48603780-48603781 on Build GRCh38
rs763752448 Chr.19: 48603809-48603809 on Build GRCh38
rs769617636 Chr.19: 48603659-48603659 on Build GRCh38
rs752385934 Chr.19: 48603712-48603712 on Build GRCh38
rs775123252 Chr.19: 48603632-48603632 on Build GRCh38
rs1016759805 Chr.19: 48603696-48603696 on Build GRCh38
rs745813433 Chr.19: 48603773-48603773 on Build GRCh38
rs946482695 Chr.19: 48603660-48603660 on Build GRCh38
rs776785961 Chr.19: 48603731-48603731 on Build GRCh38
rs757220370 Chr.19: 48603721-48603721 on Build GRCh38
rs369917515 Chr.19: 48603797-48603797 on Build GRCh38
rs754452296 Chr.19: 48603704-48603704 on Build GRCh38
rs763699204 Chr.19: 48603751-48603751 on Build GRCh38
rs186446771 Chr.19: 48603770-48603770 on Build GRCh38
rs765679519 Chr.19: 48603634-48603634 on Build GRCh38
rs560890077 Chr.19: 48603781-48603781 on Build GRCh38
rs747077302 Chr.19: 48603725-48603725 on Build GRCh38
rs540259397 Chr.19: 48603786-48603786 on Build GRCh38
rs75363654 Chr.19: 48603803-48603803 on Build GRCh38
rs771897998 Chr.19: 48603626-48603626 on Build GRCh38
rs767132202 Chr.19: 48603708-48603708 on Build GRCh38
rs766951316 Chr.19: 48603692-48603692 on Build GRCh38
rs777519268 Chr.19: 48603717-48603717 on Build GRCh38
rs767004577 Chr.19: 48603735-48603735 on Build GRCh38
rs760063320 Chr.19: 48603789-48603789 on Build GRCh38
rs776459625 Chr.19: 48603785-48603785 on Build GRCh38
rs779681846 Chr.19: 48603650-48603650 on Build GRCh38
rs765692637 Chr.19: 48603683-48603683 on Build GRCh38
rs763532814 Chr.19: 48603688-48603688 on Build GRCh38
rs777752019 Chr.19: 48603667-48603667 on Build GRCh38
rs764332188 Chr.19: 48603629-48603629 on Build GRCh38
rs778081953 Chr.19: 48603783-48603783 on Build GRCh38
rs575086876 Chr.19: 48603746-48603746 on Build GRCh38
rs749030951 Chr.19: 48603718-48603718 on Build GRCh38
rs773735468 Chr.19: 48603733-48603733 on Build GRCh38
rs577301127 Chr.19: 48603730-48603730 on Build GRCh38
rs774756725 Chr.19: 48603788-48603788 on Build GRCh38
rs748372083 Chr.19: 48603732-48603732 on Build GRCh38
rs753480035 Chr.19: 48603814-48603814 on Build GRCh38
rs201027346 Chr.19: 48603782-48603782 on Build GRCh38
rs566742854 Chr.19: 48603670-48603670 on Build GRCh38
rs763341954 Chr.19: 48603734-48603734 on Build GRCh38
rs558759855 Chr.19: 48603723-48603723 on Build GRCh38
rs778777979 Chr.19: 48603771-48603771 on Build GRCh38
rs538355053 Chr.19: 48603737-48603737 on Build GRCh38
rs753638172 Chr.19: 48603755-48603755 on Build GRCh38
rs3745728 Chr.19: 48603738-48603738 on Build GRCh38
rs887845430 Chr.19: 48603693-48603693 on Build GRCh38
rs868149081 Chr.19: 48603625-48603625 on Build GRCh38
rs761667452 Chr.19: 48603815-48603815 on Build GRCh38
rs146946394 Chr.19: 48603800-48603800 on Build GRCh38
rs775093735 Chr.19: 48603801-48603801 on Build GRCh38
rs751307477 Chr.19: 48603651-48603651 on Build GRCh38
rs780828161 Chr.19: 48603658-48603658 on Build GRCh38
rs940637755 Chr.19: 48603669-48603669 on Build GRCh38
rs938200092 Chr.19: 48603677-48603677 on Build GRCh38
rs755783157 Chr.19: 48603750-48603750 on Build GRCh38
rs775571949 Chr.19: 48603671-48603671 on Build GRCh38
rs564745836 Chr.19: 48603787-48603787 on Build GRCh38
rs762142799 Chr.19: 48603682-48603682 on Build GRCh38
rs371295065 Chr.19: 48603652-48603652 on Build GRCh38
rs750954679 Chr.19: 48603649-48603649 on Build GRCh38

View Full Product Details