Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42830013-42830268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTSCR1L
Entrez Gene ID
23506
Gene Name
GLTSCR1 like
Gene Aliases
KIAA0240
UniGene
Hs.658033
Gene Chromosome Location
Chr.6: 42746958-42868560 on Build GRCh38
Transcripts
NM_001318819.1 NM_015349.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3430681 c.1830C>A p.F610L Chr.6: 42830163-42830163 on Build GRCh38
5559764 c.1712T>C p.L571P Chr.6: 42830045-42830045 on Build GRCh38
6671877 c.1772C>A p.S591Y Chr.6: 42830105-42830105 on Build GRCh38
4153408 c.1698T>C p.F566F Chr.6: 42830031-42830031 on Build GRCh38
5925283 c.1839+8C>T p.? Chr.6: 42830180-42830180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs193921035 Chr.6: 42830111-42830111 on Build GRCh38
rs753611286 Chr.6: 42830226-42830226 on Build GRCh38
rs774334662 Chr.6: 42830071-42830071 on Build GRCh38
rs370028249 Chr.6: 42830176-42830176 on Build GRCh38
rs766092333 Chr.6: 42830200-42830200 on Build GRCh38
rs149171648 Chr.6: 42830114-42830114 on Build GRCh38
rs760141598 Chr.6: 42830161-42830163 on Build GRCh38
rs761866906 Chr.6: 42830188-42830188 on Build GRCh38
rs774907499 Chr.6: 42830186-42830186 on Build GRCh38
rs757918730 Chr.6: 42830134-42830134 on Build GRCh38
rs529305180 Chr.6: 42830208-42830208 on Build GRCh38
rs754908273 Chr.6: 42830108-42830108 on Build GRCh38
rs767908263 Chr.6: 42830085-42830085 on Build GRCh38
rs899315164 Chr.6: 42830126-42830126 on Build GRCh38
rs753620351 Chr.6: 42830225-42830225 on Build GRCh38
rs780427707 Chr.6: 42830171-42830171 on Build GRCh38
rs765148566 Chr.6: 42830229-42830229 on Build GRCh38
rs753621216 Chr.6: 42830094-42830094 on Build GRCh38
rs777194999 Chr.6: 42830135-42830135 on Build GRCh38
rs758271154 Chr.6: 42830235-42830235 on Build GRCh38
rs752682002 Chr.6: 42830230-42830230 on Build GRCh38
rs887110854 Chr.6: 42830109-42830109 on Build GRCh38
rs372222168 Chr.6: 42830217-42830217 on Build GRCh38
rs766290946 Chr.6: 42830090-42830090 on Build GRCh38
rs765596531 Chr.6: 42830188-42830191 on Build GRCh38
rs763388560 Chr.6: 42830044-42830044 on Build GRCh38
rs770518912 Chr.6: 42830164-42830164 on Build GRCh38
rs769090789 Chr.6: 42830184-42830184 on Build GRCh38
rs769165870 Chr.6: 42830061-42830061 on Build GRCh38
rs752201203 Chr.6: 42830131-42830131 on Build GRCh38
rs772284688 Chr.6: 42830197-42830197 on Build GRCh38
rs144992246 Chr.6: 42830089-42830089 on Build GRCh38
rs143301030 Chr.6: 42830145-42830145 on Build GRCh38
rs1037911450 Chr.6: 42830117-42830117 on Build GRCh38
rs761929434 Chr.6: 42830073-42830073 on Build GRCh38
rs750671253 Chr.6: 42830086-42830086 on Build GRCh38
rs34239227 Chr.6: 42830224-42830225 on Build GRCh38

View Full Product Details