Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161124089-161124320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEDD
Entrez Gene ID
9191
Gene Name
death effector domain containing
Gene Aliases
CASP8IP1, DEDD1, DEFT, FLDED1, KE05
UniGene
Hs.744092
Gene Chromosome Location
Chr.1: 161120977-161132783 on Build GRCh38
Transcripts
NM_001039711.1 NM_001039712.1 NM_032998.2 XM_005245597.3 XM_005245598.4 XM_005245599.2 XM_005245600.2 XM_017002802.1 NM_001185092.1 XM_006711341.3 XM_017001371.1

Gene Symbol
NIT1
Entrez Gene ID
4817
Gene Name
nitrilase 1
Gene Aliases
-
UniGene
Hs.146406
Gene Chromosome Location
Chr.1: 161118072-161125446 on Build GRCh38
Transcripts
NM_001039711.1 NM_001039712.1 NM_032998.2 XM_005245597.3 XM_005245598.4 XM_005245599.2 XM_005245600.2 XM_017002802.1 NM_001185092.1 XM_006711341.3 XM_017001371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5508266 c.173G>A p.R58H Chr.1: 161124290-161124290 on Build GRCh38
4426388 c.252G>A p.Q84Q Chr.1: 161124211-161124211 on Build GRCh38
228277 c.319C>T p.R107W Chr.1: 161124144-161124144 on Build GRCh38
6059060 c.247C>T p.R83C Chr.1: 161124216-161124216 on Build GRCh38
898473 c.219C>T p.R73R Chr.1: 161124244-161124244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867247856 Chr.1: 161124245-161124249 on Build GRCh38
rs755999298 Chr.1: 161124237-161124237 on Build GRCh38
rs777202526 Chr.1: 161124194-161124194 on Build GRCh38
rs868587185 Chr.1: 161124252-161124252 on Build GRCh38
rs748771242 Chr.1: 161124245-161124245 on Build GRCh38
rs769362055 Chr.1: 161124157-161124157 on Build GRCh38
rs754825051 Chr.1: 161124229-161124229 on Build GRCh38
rs375230578 Chr.1: 161124118-161124118 on Build GRCh38
rs776848050 Chr.1: 161124286-161124286 on Build GRCh38
rs751518775 Chr.1: 161124213-161124213 on Build GRCh38
rs529411793 Chr.1: 161124205-161124205 on Build GRCh38
rs748758614 Chr.1: 161124290-161124290 on Build GRCh38
rs139874657 Chr.1: 161124144-161124144 on Build GRCh38
rs762845334 Chr.1: 161124110-161124110 on Build GRCh38
rs778571118 Chr.1: 161124261-161124261 on Build GRCh38
rs200035776 Chr.1: 161124183-161124183 on Build GRCh38
rs933289412 Chr.1: 161124292-161124292 on Build GRCh38
rs773422609 Chr.1: 161124215-161124215 on Build GRCh38
rs769081971 Chr.1: 161124270-161124270 on Build GRCh38
rs770537234 Chr.1: 161124174-161124174 on Build GRCh38
rs762171480 Chr.1: 161124201-161124201 on Build GRCh38
rs754848698 Chr.1: 161124216-161124216 on Build GRCh38
rs780060038 Chr.1: 161124114-161124114 on Build GRCh38
rs777379767 Chr.1: 161124244-161124244 on Build GRCh38
rs781221087 Chr.1: 161124143-161124143 on Build GRCh38
rs145580342 Chr.1: 161124163-161124163 on Build GRCh38
rs751790653 Chr.1: 161124217-161124217 on Build GRCh38
rs563739629 Chr.1: 161124181-161124181 on Build GRCh38
rs752735474 Chr.1: 161124235-161124235 on Build GRCh38
rs775878319 Chr.1: 161124178-161124178 on Build GRCh38
rs151010008 Chr.1: 161124246-161124246 on Build GRCh38
rs754771875 Chr.1: 161124132-161124132 on Build GRCh38
rs766121975 Chr.1: 161124180-161124180 on Build GRCh38
rs770170298 Chr.1: 161124295-161124295 on Build GRCh38
rs767158548 Chr.1: 161124230-161124230 on Build GRCh38
rs576987517 Chr.1: 161124123-161124123 on Build GRCh38
rs745459751 Chr.1: 161124269-161124269 on Build GRCh38
rs974610635 Chr.1: 161124203-161124203 on Build GRCh38
rs777136200 Chr.1: 161124162-161124162 on Build GRCh38

View Full Product Details