Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 170317651-170317916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120B
Entrez Gene ID
84498
Gene Name
family with sequence similarity 120B
Gene Aliases
CCPG, KIAA1838, PGCC1, dJ894D12.1
UniGene
Hs.369522
Gene Chromosome Location
Chr.6: 170290703-170407748 on Build GRCh38
Transcripts
NM_001286379.1 NM_001286380.1 NM_001286381.1 NM_032448.2 XM_011536185.2 XM_017011358.1 XM_017011359.1 XM_017011360.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4353058 c.384G>A p.R128R Chr.6: 170317705-170317705 on Build GRCh38
6173605 c.471C>T p.P157P Chr.6: 170317792-170317792 on Build GRCh38
4353059 c.384G>C p.R128S Chr.6: 170317705-170317705 on Build GRCh38
7327213 c.498T>G p.Y166* Chr.6: 170317888-170317888 on Build GRCh38
4353056 c.333C>A p.F111L Chr.6: 170317654-170317654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779533323 Chr.6: 170317887-170317887 on Build GRCh38
rs761896291 Chr.6: 170317759-170317759 on Build GRCh38
rs149247236 Chr.6: 170317818-170317818 on Build GRCh38
rs867802554 Chr.6: 170317769-170317769 on Build GRCh38
rs780725538 Chr.6: 170317865-170317865 on Build GRCh38
rs778762247 Chr.6: 170317686-170317686 on Build GRCh38
rs771356096 Chr.6: 170317817-170317817 on Build GRCh38
rs775905378 Chr.6: 170317729-170317729 on Build GRCh38
rs758384293 Chr.6: 170317694-170317694 on Build GRCh38
rs372177611 Chr.6: 170317676-170317676 on Build GRCh38
rs752555518 Chr.6: 170317834-170317834 on Build GRCh38
rs3800266 Chr.6: 170317855-170317855 on Build GRCh38
rs904966764 Chr.6: 170317727-170317727 on Build GRCh38
rs748076725 Chr.6: 170317692-170317692 on Build GRCh38
rs769513162 Chr.6: 170317877-170317877 on Build GRCh38
rs200797523 Chr.6: 170317758-170317758 on Build GRCh38
rs758222570 Chr.6: 170317842-170317842 on Build GRCh38
rs770496011 Chr.6: 170317707-170317707 on Build GRCh38
rs779176351 Chr.6: 170317839-170317839 on Build GRCh38
rs758435301 Chr.6: 170317845-170317846 on Build GRCh38
rs895674425 Chr.6: 170317741-170317741 on Build GRCh38
rs201436299 Chr.6: 170317677-170317677 on Build GRCh38
rs749658778 Chr.6: 170317890-170317890 on Build GRCh38
rs138355463 Chr.6: 170317768-170317768 on Build GRCh38
rs764989085 Chr.6: 170317802-170317802 on Build GRCh38
rs760919116 Chr.6: 170317801-170317801 on Build GRCh38
rs776487833 Chr.6: 170317709-170317709 on Build GRCh38
rs767569996 Chr.6: 170317763-170317763 on Build GRCh38
rs754785962 Chr.6: 170317684-170317684 on Build GRCh38
rs777855786 Chr.6: 170317854-170317854 on Build GRCh38
rs1005120731 Chr.6: 170317751-170317751 on Build GRCh38
rs1002264857 Chr.6: 170317831-170317831 on Build GRCh38
rs757287763 Chr.6: 170317675-170317675 on Build GRCh38
rs746546544 Chr.6: 170317705-170317705 on Build GRCh38
rs937395904 Chr.6: 170317685-170317685 on Build GRCh38
rs745561142 Chr.6: 170317871-170317871 on Build GRCh38
rs35221221 Chr.6: 170317792-170317792 on Build GRCh38
rs763179480 Chr.6: 170317732-170317732 on Build GRCh38
rs770100452 Chr.6: 170317728-170317728 on Build GRCh38
rs745532990 Chr.6: 170317717-170317717 on Build GRCh38
rs777800651 Chr.6: 170317701-170317701 on Build GRCh38
rs764617377 Chr.6: 170317749-170317749 on Build GRCh38
rs756898165 Chr.6: 170317860-170317860 on Build GRCh38
rs747424745 Chr.6: 170317700-170317700 on Build GRCh38

View Full Product Details