Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22907776-22908033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB2
Entrez Gene ID
2048
Gene Name
EPH receptor B2
Gene Aliases
CAPB, DRT, EK5, EPHT3, ERK, Hek5, PCBC, Tyro5
UniGene
Hs.380705
Gene Chromosome Location
Chr.1: 22710770-22921500 on Build GRCh38
Transcripts
NM_001309192.1 NM_001309193.1 NM_004442.7 NM_017449.4 XM_006710441.3 XM_006710442.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
905549 c.2191G>A p.A731T Chr.1: 22908007-22908007 on Build GRCh38
7126856 c.2205G>T p.M735I Chr.1: 22908018-22908018 on Build GRCh38
7126857 c.2202G>T p.M734I Chr.1: 22908018-22908018 on Build GRCh38
1667246 c.2164C>T p.Q722* Chr.1: 22907980-22907980 on Build GRCh38
4941591 c.2143G>A p.D715N Chr.1: 22907959-22907959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750622593 Chr.1: 22907903-22907903 on Build GRCh38
rs375579646 Chr.1: 22908003-22908003 on Build GRCh38
rs773617571 Chr.1: 22907968-22907968 on Build GRCh38
rs910253794 Chr.1: 22907796-22907796 on Build GRCh38
rs142161660 Chr.1: 22908007-22908007 on Build GRCh38
rs368409603 Chr.1: 22907920-22907920 on Build GRCh38
rs751896309 Chr.1: 22907959-22907959 on Build GRCh38
rs748623989 Chr.1: 22907955-22907955 on Build GRCh38
rs1047307944 Chr.1: 22907894-22907894 on Build GRCh38
rs771816281 Chr.1: 22907999-22907999 on Build GRCh38
rs375588609 Chr.1: 22908006-22908006 on Build GRCh38
rs1005191803 Chr.1: 22907927-22907927 on Build GRCh38
rs201544391 Chr.1: 22907976-22907976 on Build GRCh38
rs896021035 Chr.1: 22907838-22907838 on Build GRCh38
rs753027458 Chr.1: 22907935-22907936 on Build GRCh38
rs121912582 Chr.1: 22907980-22907980 on Build GRCh38
rs963824341 Chr.1: 22907997-22907997 on Build GRCh38
rs764219953 Chr.1: 22907875-22907875 on Build GRCh38
rs780148814 Chr.1: 22907925-22907925 on Build GRCh38
rs370900320 Chr.1: 22907982-22907982 on Build GRCh38
rs936888028 Chr.1: 22907950-22907950 on Build GRCh38
rs111404899 Chr.1: 22907813-22907813 on Build GRCh38
rs768675034 Chr.1: 22907929-22907929 on Build GRCh38
rs1018379423 Chr.1: 22907986-22907986 on Build GRCh38
rs754226740 Chr.1: 22907958-22907958 on Build GRCh38
rs892649210 Chr.1: 22907988-22907988 on Build GRCh38
rs781582583 Chr.1: 22907936-22907936 on Build GRCh38

View Full Product Details