Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37347113-37347386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HKR1
Entrez Gene ID
284459
Gene Name
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases
ZNF875
UniGene
Hs.657027
Gene Chromosome Location
Chr.19: 37317900-37364455 on Build GRCh38
Transcripts
NM_181786.2 XM_017026672.1 XM_017026673.1 XM_017026674.1 XM_017026675.1 XM_017026676.1 XM_017026677.1 XM_017026678.1 XM_017026679.1 XM_017026680.1 XM_017026681.1 XM_017026682.1 XM_017026683.1 XM_017026684.1 XM_017026685.1 XM_017026686.1 XM_017026687.1 XM_017026688.1 XM_017026689.1 XM_017026690.1 XM_017026691.1 XM_017026692.1 XM_017026693.1 XM_017026694.1 XM_017026695.1 XM_017026696.1 XM_017026697.1 XM_017026698.1 XM_017026699.1 XM_017026700.1 XM_017026701.1 XM_017026702.1 XM_017026703.1 XM_017026704.1 XM_017026706.1 XM_017026707.1 XM_017026708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4282292 c.129G>C p.M43I Chr.19: 37347285-37347285 on Build GRCh38
188406 c.96C>T p.F32F Chr.19: 37347195-37347195 on Build GRCh38
4282290 c.40G>A p.V14M Chr.19: 37347196-37347196 on Build GRCh38
3787655 c.170T>C p.L57P Chr.19: 37347269-37347269 on Build GRCh38
5963669 c.183G>T p.V61V Chr.19: 37347282-37347282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748966240 Chr.19: 37347319-37347319 on Build GRCh38
rs781382773 Chr.19: 37347157-37347157 on Build GRCh38
rs35388283 Chr.19: 37347220-37347220 on Build GRCh38
rs142786854 Chr.19: 37347222-37347222 on Build GRCh38
rs750597615 Chr.19: 37347274-37347274 on Build GRCh38
rs549724058 Chr.19: 37347199-37347199 on Build GRCh38
rs749094008 Chr.19: 37347195-37347195 on Build GRCh38
rs761768102 Chr.19: 37347207-37347207 on Build GRCh38
rs751993604 Chr.19: 37347313-37347313 on Build GRCh38
rs770179132 Chr.19: 37347191-37347191 on Build GRCh38
rs770370006 Chr.19: 37347302-37347302 on Build GRCh38
rs774156728 Chr.19: 37347196-37347196 on Build GRCh38
rs750902219 Chr.19: 37347348-37347348 on Build GRCh38
rs745919867 Chr.19: 37347172-37347172 on Build GRCh38
rs900732670 Chr.19: 37347137-37347137 on Build GRCh38
rs771006012 Chr.19: 37347295-37347295 on Build GRCh38
rs747228366 Chr.19: 37347228-37347228 on Build GRCh38
rs756250183 Chr.19: 37347275-37347275 on Build GRCh38
rs749457125 Chr.19: 37347279-37347279 on Build GRCh38
rs758788780 Chr.19: 37347141-37347141 on Build GRCh38
rs776153383 Chr.19: 37347303-37347303 on Build GRCh38
rs759190495 Chr.19: 37347304-37347304 on Build GRCh38
rs915202014 Chr.19: 37347332-37347332 on Build GRCh38
rs768365093 Chr.19: 37347287-37347287 on Build GRCh38
rs371006116 Chr.19: 37347215-37347215 on Build GRCh38
rs752039587 Chr.19: 37347235-37347235 on Build GRCh38
rs767034321 Chr.19: 37347214-37347214 on Build GRCh38
rs990785973 Chr.19: 37347331-37347331 on Build GRCh38
rs757791297 Chr.19: 37347239-37347239 on Build GRCh38
rs141089018 Chr.19: 37347297-37347297 on Build GRCh38
rs115422824 Chr.19: 37347309-37347309 on Build GRCh38
rs751253429 Chr.19: 37347330-37347330 on Build GRCh38
rs780149681 Chr.19: 37347277-37347277 on Build GRCh38
rs775851120 Chr.19: 37347192-37347192 on Build GRCh38
rs932722554 Chr.19: 37347355-37347355 on Build GRCh38
rs151129355 Chr.19: 37347231-37347231 on Build GRCh38
rs377692121 Chr.19: 37347146-37347146 on Build GRCh38
rs757418754 Chr.19: 37347148-37347148 on Build GRCh38
rs753519959 Chr.19: 37347227-37347227 on Build GRCh38
rs778718963 Chr.19: 37347294-37347294 on Build GRCh38
rs781754224 Chr.19: 37347271-37347271 on Build GRCh38
rs1015455630 Chr.19: 37347282-37347282 on Build GRCh38
rs376328994 Chr.19: 37347326-37347326 on Build GRCh38
rs1042423287 Chr.19: 37347361-37347361 on Build GRCh38

View Full Product Details