Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128166832-128167099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIZ1
Entrez Gene ID
25792
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
LSFR1, NP94, ZNF356
UniGene
Hs.212395
Gene Chromosome Location
Chr.9: 128166065-128204383 on Build GRCh38
Transcripts
NM_001131015.1 NM_001131016.1 NM_001131017.1 NM_001131018.1 NM_001257975.1 NM_001257976.1 NM_012127.2 XM_005251888.3 XM_005251891.3 XM_005251892.3 XM_005251893.4 XM_006717039.3 XM_011518481.1 XM_011518482.2 XM_011518483.1 XM_017014594.1 XM_017014595.1 XM_017014596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1736465 c.2404C>T p.R802C Chr.9: 128166842-128166842 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs536128667 Chr.9: 128166965-128166965 on Build GRCh38
rs368596460 Chr.9: 128166891-128166891 on Build GRCh38
rs878888388 Chr.9: 128167003-128167003 on Build GRCh38
rs15126 Chr.9: 128166877-128166877 on Build GRCh38
rs200887788 Chr.9: 128167047-128167047 on Build GRCh38
rs893461875 Chr.9: 128166873-128166873 on Build GRCh38
rs889517118 Chr.9: 128166989-128166989 on Build GRCh38
rs954919848 Chr.9: 128166950-128166950 on Build GRCh38
rs1049500506 Chr.9: 128166988-128166988 on Build GRCh38
rs181229235 Chr.9: 128166967-128166967 on Build GRCh38
rs1019253511 Chr.9: 128167031-128167031 on Build GRCh38
rs374054563 Chr.9: 128166879-128166879 on Build GRCh38
rs757675037 Chr.9: 128166888-128166889 on Build GRCh38
rs150886477 Chr.9: 128166945-128166945 on Build GRCh38
rs775531056 Chr.9: 128166857-128166857 on Build GRCh38
rs73672437 Chr.9: 128166915-128166915 on Build GRCh38
rs757596071 Chr.9: 128166871-128166871 on Build GRCh38
rs575531119 Chr.9: 128166978-128166978 on Build GRCh38
rs751886503 Chr.9: 128166863-128166863 on Build GRCh38
rs544225182 Chr.9: 128167025-128167025 on Build GRCh38
rs762965441 Chr.9: 128167064-128167064 on Build GRCh38
rs139673040 Chr.9: 128167038-128167038 on Build GRCh38
rs747658553 Chr.9: 128166906-128166906 on Build GRCh38
rs750980260 Chr.9: 128166887-128166887 on Build GRCh38
rs778625485 Chr.9: 128166895-128166895 on Build GRCh38
rs189607151 Chr.9: 128166926-128166926 on Build GRCh38
rs540341207 Chr.9: 128167058-128167058 on Build GRCh38
rs185085914 Chr.9: 128166861-128166861 on Build GRCh38
rs541044005 Chr.9: 128166864-128166864 on Build GRCh38
rs201948497 Chr.9: 128166927-128166927 on Build GRCh38
rs745859418 Chr.9: 128167056-128167056 on Build GRCh38
rs750628165 Chr.9: 128166974-128166974 on Build GRCh38
rs140324491 Chr.9: 128166860-128166860 on Build GRCh38
rs777433795 Chr.9: 128166918-128166918 on Build GRCh38

View Full Product Details