Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70659942-70660215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL34
Entrez Gene ID
146433
Gene Name
interleukin 34
Gene Aliases
C16orf77, IL-34
UniGene
Hs.461214
Gene Chromosome Location
Chr.16: 70579895-70660682 on Build GRCh38
Transcripts
NM_001172771.1 NM_001172772.1 NM_152456.2 XM_011522902.2 XM_011522903.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
417291 c.686C>T p.S229L Chr.16: 70660144-70660144 on Build GRCh38
6685336 c.552C>T p.S184S Chr.16: 70660010-70660010 on Build GRCh38
417292 c.623C>T p.A208V Chr.16: 70660081-70660081 on Build GRCh38
5971897 c.659C>A p.P220H Chr.16: 70660117-70660117 on Build GRCh38
4062388 c.618G>C p.Q206H Chr.16: 70660076-70660076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048635773 Chr.16: 70660162-70660162 on Build GRCh38
rs753771953 Chr.16: 70660158-70660158 on Build GRCh38
rs4985556 Chr.16: 70660097-70660097 on Build GRCh38
rs774449875 Chr.16: 70660002-70660002 on Build GRCh38
rs370957526 Chr.16: 70660194-70660194 on Build GRCh38
rs754912577 Chr.16: 70660164-70660164 on Build GRCh38
rs927551998 Chr.16: 70659962-70659962 on Build GRCh38
rs746523737 Chr.16: 70660163-70660164 on Build GRCh38
rs551796755 Chr.16: 70660183-70660183 on Build GRCh38
rs7206509 Chr.16: 70660042-70660042 on Build GRCh38
rs201952472 Chr.16: 70659981-70659981 on Build GRCh38
rs768715713 Chr.16: 70659993-70659993 on Build GRCh38
rs745769402 Chr.16: 70660078-70660078 on Build GRCh38
rs755937255 Chr.16: 70660112-70660112 on Build GRCh38
rs766224487 Chr.16: 70660154-70660154 on Build GRCh38
rs747139320 Chr.16: 70659969-70659970 on Build GRCh38
rs750167732 Chr.16: 70660111-70660111 on Build GRCh38
rs780602039 Chr.16: 70660175-70660175 on Build GRCh38
rs746167704 Chr.16: 70660131-70660131 on Build GRCh38
rs3214847 Chr.16: 70659970-70659971 on Build GRCh38
rs746149647 Chr.16: 70660171-70660171 on Build GRCh38
rs779902217 Chr.16: 70660113-70660113 on Build GRCh38
rs761996710 Chr.16: 70660006-70660006 on Build GRCh38
rs747223979 Chr.16: 70660126-70660126 on Build GRCh38
rs397711055 Chr.16: 70659980-70659981 on Build GRCh38
rs772052353 Chr.16: 70660087-70660087 on Build GRCh38
rs141513638 Chr.16: 70660153-70660153 on Build GRCh38
rs777183793 Chr.16: 70660045-70660045 on Build GRCh38
rs771592770 Chr.16: 70659985-70659986 on Build GRCh38
rs144464320 Chr.16: 70660173-70660173 on Build GRCh38
rs200144613 Chr.16: 70660109-70660109 on Build GRCh38
rs769403976 Chr.16: 70660094-70660094 on Build GRCh38
rs752305128 Chr.16: 70660057-70660057 on Build GRCh38
rs150579141 Chr.16: 70660169-70660169 on Build GRCh38
rs773778670 Chr.16: 70660144-70660144 on Build GRCh38
rs200444183 Chr.16: 70660139-70660139 on Build GRCh38
rs577167294 Chr.16: 70659967-70659967 on Build GRCh38
rs755146539 Chr.16: 70659964-70659964 on Build GRCh38
rs781232520 Chr.16: 70660147-70660147 on Build GRCh38
rs758011880 Chr.16: 70660062-70660062 on Build GRCh38
rs768176846 Chr.16: 70660142-70660142 on Build GRCh38
rs748188942 Chr.16: 70660083-70660083 on Build GRCh38
rs780153224 Chr.16: 70659996-70659996 on Build GRCh38
rs762466829 Chr.16: 70660103-70660103 on Build GRCh38
rs576160948 Chr.16: 70659970-70659970 on Build GRCh38
rs570123541 Chr.16: 70660184-70660184 on Build GRCh38
rs372998917 Chr.16: 70660138-70660138 on Build GRCh38
rs777016133 Chr.16: 70660129-70660129 on Build GRCh38
rs189265997 Chr.16: 70659980-70659980 on Build GRCh38
rs747715186 Chr.16: 70659988-70659988 on Build GRCh38
rs369568962 Chr.16: 70660034-70660034 on Build GRCh38
rs757060030 Chr.16: 70660072-70660072 on Build GRCh38
rs138439611 Chr.16: 70660088-70660088 on Build GRCh38
rs745592299 Chr.16: 70659983-70659984 on Build GRCh38
rs202122982 Chr.16: 70660098-70660098 on Build GRCh38
rs749776814 Chr.16: 70660182-70660182 on Build GRCh38
rs201784459 Chr.16: 70660102-70660102 on Build GRCh38
rs201277640 Chr.16: 70660081-70660081 on Build GRCh38
rs148286339 Chr.16: 70660108-70660108 on Build GRCh38
rs79922158 Chr.16: 70660067-70660067 on Build GRCh38
rs777887915 Chr.16: 70660167-70660167 on Build GRCh38
rs772292258 Chr.16: 70660010-70660010 on Build GRCh38
rs192356278 Chr.16: 70660145-70660145 on Build GRCh38
rs764810390 Chr.16: 70660055-70660055 on Build GRCh38
rs112639369 Chr.16: 70660170-70660170 on Build GRCh38
rs751216629 Chr.16: 70660107-70660107 on Build GRCh38
rs749242332 Chr.16: 70660114-70660114 on Build GRCh38
rs199910768 Chr.16: 70660082-70660082 on Build GRCh38
rs763733561 Chr.16: 70660063-70660063 on Build GRCh38
rs779361515 Chr.16: 70659981-70659981 on Build GRCh38
rs775115913 Chr.16: 70660041-70660042 on Build GRCh38
rs889733429 Chr.16: 70660105-70660105 on Build GRCh38
rs532486484 Chr.16: 70660117-70660117 on Build GRCh38
rs761163867 Chr.16: 70660191-70660191 on Build GRCh38
rs145782768 Chr.16: 70660076-70660076 on Build GRCh38
rs773341434 Chr.16: 70660011-70660011 on Build GRCh38
rs796937232 Chr.16: 70659970-70659971 on Build GRCh38
rs763570329 Chr.16: 70660106-70660106 on Build GRCh38
rs550608624 Chr.16: 70660127-70660127 on Build GRCh38

View Full Product Details