Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38727158-38727419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP1
Entrez Gene ID
9567
Gene Name
GTP binding protein 1
Gene Aliases
GP-1, GP1, HSPC018
UniGene
Hs.276925
Gene Chromosome Location
Chr.22: 38704561-38738265 on Build GRCh38
Transcripts
NM_004286.4 XM_011530537.2 XM_017029099.1 XM_017029100.1 XM_017029101.1 XM_017029102.1 XM_017029103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6442989 c.1519C>T p.P507S Chr.22: 38727330-38727330 on Build GRCh38
3554426 c.174T>G p.V58V Chr.22: 38727251-38727251 on Build GRCh38
5768973 c.1487T>C p.I496T Chr.22: 38727298-38727298 on Build GRCh38
5742118 c.1483G>A p.E495K Chr.22: 38727294-38727294 on Build GRCh38
5768974 c.221T>C p.I74T Chr.22: 38727298-38727298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756767282 Chr.22: 38727397-38727397 on Build GRCh38
rs761988740 Chr.22: 38727333-38727333 on Build GRCh38
rs915902323 Chr.22: 38727300-38727300 on Build GRCh38
rs555608974 Chr.22: 38727367-38727367 on Build GRCh38
rs945760322 Chr.22: 38727294-38727294 on Build GRCh38
rs757367839 Chr.22: 38727231-38727231 on Build GRCh38
rs369281526 Chr.22: 38727259-38727259 on Build GRCh38
rs537370099 Chr.22: 38727338-38727338 on Build GRCh38
rs759158653 Chr.22: 38727303-38727303 on Build GRCh38
rs753815957 Chr.22: 38727332-38727332 on Build GRCh38
rs751927632 Chr.22: 38727383-38727383 on Build GRCh38
rs753953156 Chr.22: 38727224-38727224 on Build GRCh38
rs372637758 Chr.22: 38727331-38727331 on Build GRCh38
rs768690159 Chr.22: 38727293-38727293 on Build GRCh38
rs760618071 Chr.22: 38727324-38727324 on Build GRCh38
rs1041354863 Chr.22: 38727320-38727320 on Build GRCh38
rs750715907 Chr.22: 38727254-38727254 on Build GRCh38
rs766696315 Chr.22: 38727277-38727277 on Build GRCh38
rs938435064 Chr.22: 38727359-38727359 on Build GRCh38
rs34233333 Chr.22: 38727395-38727395 on Build GRCh38
rs780395009 Chr.22: 38727375-38727375 on Build GRCh38
rs779222824 Chr.22: 38727236-38727236 on Build GRCh38
rs187738729 Chr.22: 38727386-38727386 on Build GRCh38
rs375582502 Chr.22: 38727385-38727385 on Build GRCh38
rs765275240 Chr.22: 38727334-38727334 on Build GRCh38
rs1055434900 Chr.22: 38727382-38727382 on Build GRCh38
rs777198868 Chr.22: 38727273-38727273 on Build GRCh38
rs922104284 Chr.22: 38727190-38727190 on Build GRCh38
rs950303152 Chr.22: 38727377-38727377 on Build GRCh38
rs990434003 Chr.22: 38727290-38727290 on Build GRCh38
rs774021549 Chr.22: 38727302-38727302 on Build GRCh38
rs758734440 Chr.22: 38727258-38727258 on Build GRCh38
rs771976065 Chr.22: 38727305-38727305 on Build GRCh38
rs769218556 Chr.22: 38727266-38727266 on Build GRCh38
rs147738144 Chr.22: 38727240-38727240 on Build GRCh38
rs775489729 Chr.22: 38727322-38727322 on Build GRCh38
rs764260498 Chr.22: 38727188-38727188 on Build GRCh38

View Full Product Details