Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24606996-24607243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZMH
Entrez Gene ID
2999
Gene Name
granzyme H
Gene Aliases
CCP-X, CGL-2, CSP-C, CTLA1, CTSGL2
UniGene
Hs.348264
Gene Chromosome Location
Chr.14: 24606480-24609763 on Build GRCh38
Transcripts
NM_001270780.1 NM_001270781.1 NM_033423.4 XM_011536683.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
137611 c.597+1G>A p.? Chr.14: 24607148-24607148 on Build GRCh38
4774182 c.590G>A p.G197D Chr.14: 24607156-24607156 on Build GRCh38
5847990 c.533G>A p.G178D Chr.14: 24607213-24607213 on Build GRCh38
325470 c.529C>A p.H177N Chr.14: 24607217-24607217 on Build GRCh38
5740064 c.583C>T p.Q195* Chr.14: 24607163-24607163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751202637 Chr.14: 24607158-24607158 on Build GRCh38
rs772350892 Chr.14: 24607102-24607102 on Build GRCh38
rs905757152 Chr.14: 24607047-24607047 on Build GRCh38
rs777224168 Chr.14: 24607132-24607132 on Build GRCh38
rs1032875873 Chr.14: 24607086-24607086 on Build GRCh38
rs745934038 Chr.14: 24607191-24607191 on Build GRCh38
rs190749747 Chr.14: 24607097-24607097 on Build GRCh38
rs373891704 Chr.14: 24607157-24607157 on Build GRCh38
rs775889211 Chr.14: 24607202-24607202 on Build GRCh38
rs77506069 Chr.14: 24607126-24607126 on Build GRCh38
rs952358122 Chr.14: 24607202-24607202 on Build GRCh38
rs1044720811 Chr.14: 24607030-24607030 on Build GRCh38
rs139103808 Chr.14: 24607196-24607196 on Build GRCh38
rs756039143 Chr.14: 24607179-24607179 on Build GRCh38
rs531293599 Chr.14: 24607058-24607058 on Build GRCh38
rs1015032161 Chr.14: 24607052-24607052 on Build GRCh38
rs747212291 Chr.14: 24607224-24607224 on Build GRCh38
rs780977697 Chr.14: 24607162-24607162 on Build GRCh38
rs200088850 Chr.14: 24607119-24607119 on Build GRCh38
rs769002213 Chr.14: 24607225-24607225 on Build GRCh38
rs145997482 Chr.14: 24607149-24607149 on Build GRCh38
rs886720418 Chr.14: 24607168-24607168 on Build GRCh38
rs762415361 Chr.14: 24607143-24607143 on Build GRCh38
rs75249489 Chr.14: 24607028-24607028 on Build GRCh38
rs757895960 Chr.14: 24607156-24607156 on Build GRCh38
rs201149975 Chr.14: 24607110-24607110 on Build GRCh38
rs927817516 Chr.14: 24607098-24607098 on Build GRCh38
rs775618039 Chr.14: 24607115-24607115 on Build GRCh38
rs775736011 Chr.14: 24607215-24607215 on Build GRCh38
rs764391932 Chr.14: 24607131-24607131 on Build GRCh38
rs544748518 Chr.14: 24607206-24607206 on Build GRCh38
rs748009130 Chr.14: 24607166-24607166 on Build GRCh38
rs370343845 Chr.14: 24607197-24607197 on Build GRCh38
rs765913156 Chr.14: 24607145-24607145 on Build GRCh38
rs1018612898 Chr.14: 24607124-24607124 on Build GRCh38
rs758058137 Chr.14: 24607129-24607129 on Build GRCh38
rs777741862 Chr.14: 24607184-24607184 on Build GRCh38
rs375085105 Chr.14: 24607134-24607134 on Build GRCh38

View Full Product Details