Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 93216813-93217038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRID2
Entrez Gene ID
2895
Gene Name
glutamate ionotropic receptor delta type subunit 2
Gene Aliases
GluD2, SCAR18
UniGene
Hs.162727
Gene Chromosome Location
Chr.4: 92304249-93810330 on Build GRCh38
Transcripts
NM_001286838.1 NM_001510.3 XM_011531893.2 XM_011531894.2 XM_011531895.2 XM_017008118.1 XM_017008119.1 XM_017008120.1 XM_017008121.1 XM_017008122.1 XM_017008123.1 XM_017008124.1 XM_017008125.1 XM_017008127.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
79939 c.891G>T p.R297R Chr.4: 93216839-93216839 on Build GRCh38
4391951 c.940G>T p.D314Y Chr.4: 93216888-93216888 on Build GRCh38
6101151 c.898C>T p.R300C Chr.4: 93216846-93216846 on Build GRCh38
3607054 c.890G>A p.R297Q Chr.4: 93216838-93216838 on Build GRCh38
298234 c.937A>C p.K313Q Chr.4: 93216885-93216885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561743155 Chr.4: 93216838-93216838 on Build GRCh38
rs765043362 Chr.4: 93216995-93216995 on Build GRCh38
rs944780089 Chr.4: 93216955-93216955 on Build GRCh38
rs775585992 Chr.4: 93216910-93216910 on Build GRCh38
rs761598577 Chr.4: 93216932-93216932 on Build GRCh38
rs751478717 Chr.4: 93216931-93216931 on Build GRCh38
rs747359105 Chr.4: 93216859-93216859 on Build GRCh38
rs538573274 Chr.4: 93216898-93216898 on Build GRCh38
rs901019889 Chr.4: 93217007-93217007 on Build GRCh38
rs762852368 Chr.4: 93216919-93216919 on Build GRCh38
rs745533496 Chr.4: 93216846-93216846 on Build GRCh38
rs755873386 Chr.4: 93216948-93216948 on Build GRCh38
rs746783316 Chr.4: 93216986-93216986 on Build GRCh38
rs182789669 Chr.4: 93216853-93216853 on Build GRCh38
rs772686060 Chr.4: 93216850-93216850 on Build GRCh38
rs1050384061 Chr.4: 93216873-93216873 on Build GRCh38
rs776854733 Chr.4: 93216874-93216874 on Build GRCh38
rs187713184 Chr.4: 93216894-93216894 on Build GRCh38
rs34958992 Chr.4: 93216901-93216902 on Build GRCh38
rs748637553 Chr.4: 93216849-93216849 on Build GRCh38
rs759811681 Chr.4: 93216879-93216879 on Build GRCh38
rs753351208 Chr.4: 93216960-93216960 on Build GRCh38
rs778167783 Chr.4: 93216891-93216891 on Build GRCh38
rs370367152 Chr.4: 93216907-93216907 on Build GRCh38
rs1037668863 Chr.4: 93216972-93216972 on Build GRCh38
rs779690113 Chr.4: 93216951-93216951 on Build GRCh38
rs771206137 Chr.4: 93216866-93216866 on Build GRCh38
rs764041084 Chr.4: 93216927-93216927 on Build GRCh38
rs201635818 Chr.4: 93216847-93216847 on Build GRCh38
rs374827763 Chr.4: 93216944-93216944 on Build GRCh38
rs767277237 Chr.4: 93216941-93216941 on Build GRCh38

View Full Product Details