Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209611715-209611976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK1G
Entrez Gene ID
57172
Gene Name
calcium/calmodulin dependent protein kinase IG
Gene Aliases
CLICK3, CLICKIII, VWS1, dJ272L16.1
UniGene
Hs.199068
Gene Chromosome Location
Chr.1: 209583700-209613939 on Build GRCh38
Transcripts
NM_020439.2 XM_017001866.1 XM_017001867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1689769 c.1051C>T p.P351S Chr.1: 209611927-209611927 on Build GRCh38
1179541 c.1030G>T p.E344* Chr.1: 209611906-209611906 on Build GRCh38
4562984 c.951G>A p.M317I Chr.1: 209611827-209611827 on Build GRCh38
1338504 c.984C>T p.G328G Chr.1: 209611860-209611860 on Build GRCh38
3750848 c.985G>A p.V329I Chr.1: 209611861-209611861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771607754 Chr.1: 209611950-209611950 on Build GRCh38
rs773379474 Chr.1: 209611944-209611944 on Build GRCh38
rs769462883 Chr.1: 209611915-209611915 on Build GRCh38
rs145387182 Chr.1: 209611841-209611841 on Build GRCh38
rs748058729 Chr.1: 209611770-209611770 on Build GRCh38
rs370405106 Chr.1: 209611830-209611830 on Build GRCh38
rs745624571 Chr.1: 209611852-209611852 on Build GRCh38
rs764201574 Chr.1: 209611849-209611850 on Build GRCh38
rs138107739 Chr.1: 209611884-209611884 on Build GRCh38
rs746876613 Chr.1: 209611782-209611782 on Build GRCh38
rs761225607 Chr.1: 209611860-209611860 on Build GRCh38
rs11119315 Chr.1: 209611861-209611861 on Build GRCh38
rs765465610 Chr.1: 209611957-209611957 on Build GRCh38
rs761151515 Chr.1: 209611755-209611755 on Build GRCh38
rs780912545 Chr.1: 209611851-209611851 on Build GRCh38
rs752282907 Chr.1: 209611876-209611876 on Build GRCh38
rs370112791 Chr.1: 209611741-209611741 on Build GRCh38
rs568676041 Chr.1: 209611886-209611886 on Build GRCh38
rs778447398 Chr.1: 209611745-209611745 on Build GRCh38
rs776808728 Chr.1: 209611865-209611865 on Build GRCh38
rs763192311 Chr.1: 209611796-209611796 on Build GRCh38
rs777200877 Chr.1: 209611877-209611877 on Build GRCh38
rs367925085 Chr.1: 209611816-209611816 on Build GRCh38
rs770702866 Chr.1: 209611783-209611783 on Build GRCh38
rs1055555041 Chr.1: 209611921-209611921 on Build GRCh38
rs374701638 Chr.1: 209611804-209611804 on Build GRCh38
rs370632835 Chr.1: 209611778-209611778 on Build GRCh38
rs574976255 Chr.1: 209611797-209611797 on Build GRCh38
rs765011800 Chr.1: 209611869-209611869 on Build GRCh38
rs554479000 Chr.1: 209611939-209611939 on Build GRCh38
rs373639592 Chr.1: 209611943-209611943 on Build GRCh38
rs150063130 Chr.1: 209611829-209611829 on Build GRCh38
rs773106760 Chr.1: 209611779-209611779 on Build GRCh38
rs142610896 Chr.1: 209611887-209611887 on Build GRCh38
rs764099572 Chr.1: 209611801-209611801 on Build GRCh38
rs200699828 Chr.1: 209611845-209611845 on Build GRCh38
rs750952662 Chr.1: 209611817-209611817 on Build GRCh38
rs17316402 Chr.1: 209611803-209611803 on Build GRCh38
rs1050235260 Chr.1: 209611911-209611911 on Build GRCh38
rs201849586 Chr.1: 209611902-209611902 on Build GRCh38
rs760816448 Chr.1: 209611945-209611945 on Build GRCh38
rs939328850 Chr.1: 209611917-209611917 on Build GRCh38
rs147599953 Chr.1: 209611857-209611857 on Build GRCh38
rs148199770 Chr.1: 209611878-209611878 on Build GRCh38
rs768959300 Chr.1: 209611854-209611854 on Build GRCh38
rs778408111 Chr.1: 209611836-209611836 on Build GRCh38
rs756559169 Chr.1: 209611819-209611819 on Build GRCh38
rs762009520 Chr.1: 209611855-209611855 on Build GRCh38
rs757846530 Chr.1: 209611839-209611839 on Build GRCh38
rs747734197 Chr.1: 209611837-209611837 on Build GRCh38
rs138637491 Chr.1: 209611928-209611928 on Build GRCh38
rs760008213 Chr.1: 209611951-209611951 on Build GRCh38
rs772219308 Chr.1: 209611856-209611856 on Build GRCh38
rs775704789 Chr.1: 209611787-209611787 on Build GRCh38
rs779208559 Chr.1: 209611923-209611923 on Build GRCh38
rs759788204 Chr.1: 209611868-209611868 on Build GRCh38
rs898708130 Chr.1: 209611752-209611752 on Build GRCh38
rs745739420 Chr.1: 209611894-209611894 on Build GRCh38

View Full Product Details