Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138206935-138207200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGEF3
Entrez Gene ID
57221
Gene Name
ARFGEF family member 3
Gene Aliases
A7322, BIG3, C6orf92, KIAA1244, PPP1R33, dJ171N11.1
UniGene
Hs.194408
Gene Chromosome Location
Chr.6: 138161690-138344663 on Build GRCh38
Transcripts
NM_020340.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1440750 c.141G>T p.E47D Chr.6: 138207045-138207045 on Build GRCh38
6322797 c.138-9G>T p.? Chr.6: 138207033-138207033 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777494479 Chr.6: 138207015-138207016 on Build GRCh38
rs745840056 Chr.6: 138207152-138207152 on Build GRCh38
rs1028617061 Chr.6: 138207061-138207061 on Build GRCh38
rs776297410 Chr.6: 138207087-138207087 on Build GRCh38
rs1019479544 Chr.6: 138207004-138207004 on Build GRCh38
rs780978884 Chr.6: 138207154-138207154 on Build GRCh38
rs182372969 Chr.6: 138207012-138207012 on Build GRCh38
rs529568259 Chr.6: 138207150-138207150 on Build GRCh38
rs751947237 Chr.6: 138207102-138207102 on Build GRCh38
rs201019259 Chr.6: 138207042-138207042 on Build GRCh38
rs746588715 Chr.6: 138207171-138207171 on Build GRCh38
rs771734197 Chr.6: 138207039-138207039 on Build GRCh38
rs140881091 Chr.6: 138207098-138207098 on Build GRCh38
rs375801303 Chr.6: 138207120-138207120 on Build GRCh38
rs763547125 Chr.6: 138207088-138207088 on Build GRCh38
rs770915499 Chr.6: 138207013-138207013 on Build GRCh38
rs34936283 Chr.6: 138206966-138206967 on Build GRCh38
rs773647822 Chr.6: 138207063-138207063 on Build GRCh38
rs751362993 Chr.6: 138207125-138207126 on Build GRCh38
rs6916055 Chr.6: 138207174-138207174 on Build GRCh38
rs757143800 Chr.6: 138207134-138207134 on Build GRCh38
rs763449735 Chr.6: 138207065-138207065 on Build GRCh38
rs774961171 Chr.6: 138207059-138207059 on Build GRCh38
rs951055628 Chr.6: 138207064-138207064 on Build GRCh38
rs777073254 Chr.6: 138206983-138206983 on Build GRCh38
rs762085354 Chr.6: 138207031-138207031 on Build GRCh38
rs369377953 Chr.6: 138207047-138207047 on Build GRCh38
rs767955812 Chr.6: 138207138-138207138 on Build GRCh38
rs78159300 Chr.6: 138207142-138207142 on Build GRCh38
rs373066070 Chr.6: 138207067-138207067 on Build GRCh38
rs150176338 Chr.6: 138207105-138207105 on Build GRCh38
rs745703886 Chr.6: 138207032-138207032 on Build GRCh38
rs113871324 Chr.6: 138206962-138206962 on Build GRCh38
rs561404807 Chr.6: 138206967-138206967 on Build GRCh38
rs755244727 Chr.6: 138207117-138207117 on Build GRCh38
rs552191666 Chr.6: 138207145-138207145 on Build GRCh38
rs917386283 Chr.6: 138207112-138207112 on Build GRCh38
rs779907211 Chr.6: 138207168-138207168 on Build GRCh38
rs373449274 Chr.6: 138207154-138207154 on Build GRCh38
rs752794114 Chr.6: 138207140-138207140 on Build GRCh38
rs574865478 Chr.6: 138207003-138207003 on Build GRCh38

View Full Product Details