Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89152229-89152502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP7
Entrez Gene ID
388646
Gene Name
guanylate binding protein 7
Gene Aliases
GBP4L
UniGene
Hs.591450
Gene Chromosome Location
Chr.1: 89131751-89176040 on Build GRCh38
Transcripts
NM_207398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6040092 c.584C>A p.T195K Chr.1: 89152309-89152309 on Build GRCh38
4397649 c.556G>A p.E186K Chr.1: 89152337-89152337 on Build GRCh38
6304491 c.541C>T p.R181* Chr.1: 89152352-89152352 on Build GRCh38
5508531 c.625G>T p.G209C Chr.1: 89152268-89152268 on Build GRCh38
373907 c.442C>A p.L148I Chr.1: 89152451-89152451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376404063 Chr.1: 89152301-89152301 on Build GRCh38
rs761351251 Chr.1: 89152387-89152387 on Build GRCh38
rs148584394 Chr.1: 89152461-89152461 on Build GRCh38
rs764397745 Chr.1: 89152343-89152344 on Build GRCh38
rs765200010 Chr.1: 89152428-89152428 on Build GRCh38
rs140430191 Chr.1: 89152339-89152339 on Build GRCh38
rs985956890 Chr.1: 89152340-89152340 on Build GRCh38
rs200689487 Chr.1: 89152271-89152271 on Build GRCh38
rs780479381 Chr.1: 89152467-89152467 on Build GRCh38
rs144935084 Chr.1: 89152425-89152425 on Build GRCh38
rs376697657 Chr.1: 89152456-89152456 on Build GRCh38
rs770863598 Chr.1: 89152336-89152336 on Build GRCh38
rs570323777 Chr.1: 89152458-89152458 on Build GRCh38
rs369594638 Chr.1: 89152391-89152391 on Build GRCh38
rs751079397 Chr.1: 89152283-89152283 on Build GRCh38
rs760265520 Chr.1: 89152417-89152417 on Build GRCh38
rs141756411 Chr.1: 89152351-89152351 on Build GRCh38
rs370023014 Chr.1: 89152395-89152395 on Build GRCh38
rs192538374 Chr.1: 89152448-89152448 on Build GRCh38
rs533272421 Chr.1: 89152368-89152368 on Build GRCh38
rs369247714 Chr.1: 89152361-89152361 on Build GRCh38
rs372410083 Chr.1: 89152277-89152277 on Build GRCh38
rs776775591 Chr.1: 89152269-89152269 on Build GRCh38
rs764704107 Chr.1: 89152282-89152282 on Build GRCh38
rs770244482 Chr.1: 89152275-89152275 on Build GRCh38
rs760856309 Chr.1: 89152340-89152340 on Build GRCh38
rs867870028 Chr.1: 89152409-89152409 on Build GRCh38
rs750087662 Chr.1: 89152294-89152294 on Build GRCh38
rs777770450 Chr.1: 89152259-89152259 on Build GRCh38
rs771178325 Chr.1: 89152261-89152261 on Build GRCh38
rs147357899 Chr.1: 89152359-89152359 on Build GRCh38
rs758383190 Chr.1: 89152447-89152447 on Build GRCh38
rs758162329 Chr.1: 89152338-89152338 on Build GRCh38
rs138540106 Chr.1: 89152352-89152352 on Build GRCh38
rs756399418 Chr.1: 89152463-89152463 on Build GRCh38
rs750413268 Chr.1: 89152298-89152298 on Build GRCh38
rs143254761 Chr.1: 89152319-89152319 on Build GRCh38
rs772804833 Chr.1: 89152392-89152392 on Build GRCh38
rs966586072 Chr.1: 89152260-89152260 on Build GRCh38
rs767180579 Chr.1: 89152292-89152292 on Build GRCh38
rs752617999 Chr.1: 89152436-89152436 on Build GRCh38
rs781074686 Chr.1: 89152460-89152460 on Build GRCh38
rs540928843 Chr.1: 89152323-89152323 on Build GRCh38
rs754018358 Chr.1: 89152451-89152451 on Build GRCh38
rs562450376 Chr.1: 89152333-89152333 on Build GRCh38
rs201388599 Chr.1: 89152384-89152384 on Build GRCh38
rs569134617 Chr.1: 89152367-89152367 on Build GRCh38
rs374864805 Chr.1: 89152355-89152355 on Build GRCh38
rs756373704 Chr.1: 89152415-89152415 on Build GRCh38
rs754826455 Chr.1: 89152426-89152426 on Build GRCh38
rs779771451 Chr.1: 89152299-89152299 on Build GRCh38

View Full Product Details