Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 123401711-123401984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1A
Entrez Gene ID
57706
Gene Name
DENN domain containing 1A
Gene Aliases
FAM31A, KIAA1608
UniGene
Hs.744995
Gene Chromosome Location
Chr.9: 123379654-123930158 on Build GRCh38
Transcripts
NM_020946.1 NM_024820.2 XM_005252109.2 XM_005252111.4 XM_005252113.4 XM_006717195.3 XM_011518882.2 XM_011518883.2 XM_011518884.2 XM_011518885.2 XM_011518886.2 XM_011518887.2 XM_017014947.1 XM_017014948.1 XM_017014949.1 XM_017014950.1 XM_017014951.1 XM_017014952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6260336 c.1673C>A p.S558* Chr.9: 123401817-123401817 on Build GRCh38
6642252 c.1581C>T p.N527N Chr.9: 123401909-123401909 on Build GRCh38
4643313 c.1637C>T p.A546V Chr.9: 123401853-123401853 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1034952200 Chr.9: 123401904-123401904 on Build GRCh38
rs745663262 Chr.9: 123401802-123401802 on Build GRCh38
rs200740254 Chr.9: 123401921-123401921 on Build GRCh38
rs543877741 Chr.9: 123401945-123401945 on Build GRCh38
rs749262859 Chr.9: 123401798-123401798 on Build GRCh38
rs746763282 Chr.9: 123401882-123401882 on Build GRCh38
rs769339500 Chr.9: 123401914-123401914 on Build GRCh38
rs543172617 Chr.9: 123401838-123401838 on Build GRCh38
rs933113243 Chr.9: 123401960-123401960 on Build GRCh38
rs572575018 Chr.9: 123401804-123401804 on Build GRCh38
rs868412968 Chr.9: 123401853-123401853 on Build GRCh38
rs377085902 Chr.9: 123401936-123401936 on Build GRCh38
rs756241717 Chr.9: 123401791-123401791 on Build GRCh38
rs775063333 Chr.9: 123401805-123401805 on Build GRCh38
rs959457376 Chr.9: 123401813-123401813 on Build GRCh38
rs777805334 Chr.9: 123401792-123401792 on Build GRCh38
rs745805005 Chr.9: 123401864-123401864 on Build GRCh38
rs963948525 Chr.9: 123401741-123401741 on Build GRCh38
rs771813359 Chr.9: 123401867-123401867 on Build GRCh38
rs747798300 Chr.9: 123401859-123401860 on Build GRCh38
rs1003484193 Chr.9: 123401902-123401902 on Build GRCh38
rs781271516 Chr.9: 123401775-123401775 on Build GRCh38
rs756296821 Chr.9: 123401846-123401846 on Build GRCh38
rs747668890 Chr.9: 123401907-123401907 on Build GRCh38
rs375724245 Chr.9: 123401806-123401806 on Build GRCh38
rs150115399 Chr.9: 123401766-123401766 on Build GRCh38
rs761360527 Chr.9: 123401822-123401822 on Build GRCh38
rs750369834 Chr.9: 123401962-123401962 on Build GRCh38
rs371321702 Chr.9: 123401922-123401922 on Build GRCh38
rs766813189 Chr.9: 123401830-123401830 on Build GRCh38
rs947277952 Chr.9: 123401926-123401926 on Build GRCh38
rs768447960 Chr.9: 123401884-123401884 on Build GRCh38
rs776315709 Chr.9: 123401819-123401819 on Build GRCh38
rs757303108 Chr.9: 123401858-123401858 on Build GRCh38
rs982279550 Chr.9: 123401767-123401767 on Build GRCh38
rs777930412 Chr.9: 123401852-123401852 on Build GRCh38
rs779868479 Chr.9: 123401879-123401879 on Build GRCh38
rs776369789 Chr.9: 123401903-123401903 on Build GRCh38
rs765338835 Chr.9: 123401950-123401950 on Build GRCh38
rs138560936 Chr.9: 123401949-123401949 on Build GRCh38
rs752907428 Chr.9: 123401782-123401782 on Build GRCh38
rs373199541 Chr.9: 123401801-123401801 on Build GRCh38
rs993795240 Chr.9: 123401746-123401746 on Build GRCh38
rs751896556 Chr.9: 123401764-123401764 on Build GRCh38
rs560218858 Chr.9: 123401814-123401814 on Build GRCh38
rs760063622 Chr.9: 123401929-123401929 on Build GRCh38
rs753991865 Chr.9: 123401946-123401946 on Build GRCh38
rs752962665 Chr.9: 123401840-123401840 on Build GRCh38
rs1026892873 Chr.9: 123401747-123401747 on Build GRCh38
rs775862168 Chr.9: 123401935-123401935 on Build GRCh38
rs561577325 Chr.9: 123401862-123401862 on Build GRCh38
rs752042108 Chr.9: 123401837-123401837 on Build GRCh38
rs751480399 Chr.9: 123401933-123401933 on Build GRCh38

View Full Product Details