Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118350501-118350744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3G
Entrez Gene ID
917
Gene Name
CD3g molecule
Gene Aliases
CD3-GAMMA, IMD17, T3G
UniGene
Hs.2259
Gene Chromosome Location
Chr.11: 118344307-118353782 on Build GRCh38
Transcripts
NM_000073.2 XM_005271724.3 XM_006718941.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
428494 c.320G>A p.C107Y Chr.11: 118350564-118350564 on Build GRCh38
3998253 c.390T>C p.A130A Chr.11: 118350634-118350634 on Build GRCh38
923544 c.355C>A p.L119I Chr.11: 118350599-118350599 on Build GRCh38
466397 c.356T>C p.L119P Chr.11: 118350600-118350600 on Build GRCh38
6519620 c.394G>A p.G132R Chr.11: 118350638-118350638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199749190 Chr.11: 118350646-118350646 on Build GRCh38
rs199691768 Chr.11: 118350528-118350528 on Build GRCh38
rs780585252 Chr.11: 118350567-118350567 on Build GRCh38
rs781239764 Chr.11: 118350597-118350597 on Build GRCh38
rs201296802 Chr.11: 118350548-118350548 on Build GRCh38
rs760675305 Chr.11: 118350639-118350639 on Build GRCh38
rs201404283 Chr.11: 118350524-118350524 on Build GRCh38
rs1052810675 Chr.11: 118350673-118350673 on Build GRCh38
rs747479547 Chr.11: 118350570-118350570 on Build GRCh38
rs916908630 Chr.11: 118350681-118350681 on Build GRCh38
rs774233735 Chr.11: 118350615-118350615 on Build GRCh38
rs755113577 Chr.11: 118350578-118350578 on Build GRCh38
rs781006056 Chr.11: 118350635-118350635 on Build GRCh38
rs202061055 Chr.11: 118350641-118350641 on Build GRCh38
rs200691936 Chr.11: 118350678-118350678 on Build GRCh38
rs3753058 Chr.11: 118350635-118350635 on Build GRCh38
rs3753059 Chr.11: 118350634-118350634 on Build GRCh38
rs905725199 Chr.11: 118350707-118350707 on Build GRCh38
rs745602409 Chr.11: 118350616-118350616 on Build GRCh38
rs374176598 Chr.11: 118350626-118350626 on Build GRCh38
rs568937392 Chr.11: 118350702-118350702 on Build GRCh38
rs776211799 Chr.11: 118350690-118350690 on Build GRCh38
rs187746068 Chr.11: 118350694-118350694 on Build GRCh38
rs71469175 Chr.11: 118350634-118350635 on Build GRCh38
rs879337484 Chr.11: 118350634-118350635 on Build GRCh38
rs916270240 Chr.11: 118350709-118350709 on Build GRCh38
rs768220101 Chr.11: 118350632-118350633 on Build GRCh38
rs769843997 Chr.11: 118350613-118350613 on Build GRCh38
rs200717244 Chr.11: 118350708-118350708 on Build GRCh38
rs200093444 Chr.11: 118350695-118350695 on Build GRCh38
rs748419520 Chr.11: 118350605-118350605 on Build GRCh38
rs765636195 Chr.11: 118350535-118350535 on Build GRCh38
rs200405767 Chr.11: 118350539-118350539 on Build GRCh38
rs934893999 Chr.11: 118350663-118350663 on Build GRCh38
rs746607767 Chr.11: 118350608-118350617 on Build GRCh38
rs928218128 Chr.11: 118350662-118350662 on Build GRCh38
rs771971617 Chr.11: 118350625-118350625 on Build GRCh38

View Full Product Details