Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75291631-75291902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
UniGene
Hs.733835
Gene Chromosome Location
Chr.12: 75275979-75390941 on Build GRCh38
Transcripts
NM_001286547.1 NM_001286548.1 NM_001286549.1 NM_032606.3 XM_005269192.1 XM_005269194.3 XM_006719648.2 XM_006719649.2 XM_011538878.1 XM_011538886.2 XM_011538887.2 XM_011538888.1 XM_011538889.2 XM_011538890.1 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020036.1 XM_017020037.1 XM_017020038.1 XM_017020039.1 XM_017020040.1 XM_017020041.1 XM_017020042.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 XM_017020046.1 XM_017020047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4044823 c.584T>C p.V195A Chr.12: 75291761-75291761 on Build GRCh38
3417141 c.525-1G>T p.? Chr.12: 75291821-75291821 on Build GRCh38
5841898 c.546G>T p.E182D Chr.12: 75291799-75291799 on Build GRCh38
340975 c.546G>A p.E182E Chr.12: 75291799-75291799 on Build GRCh38
3417140 c.1221-1G>T p.? Chr.12: 75291821-75291821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755812020 Chr.12: 75291725-75291725 on Build GRCh38
rs775802336 Chr.12: 75291789-75291789 on Build GRCh38
rs775644009 Chr.12: 75291803-75291804 on Build GRCh38
rs200661882 Chr.12: 75291772-75291772 on Build GRCh38
rs752243101 Chr.12: 75291836-75291836 on Build GRCh38
rs774160887 Chr.12: 75291824-75291824 on Build GRCh38
rs371737610 Chr.12: 75291831-75291831 on Build GRCh38
rs763712343 Chr.12: 75291855-75291855 on Build GRCh38
rs569240737 Chr.12: 75291685-75291685 on Build GRCh38
rs796239608 Chr.12: 75291830-75291830 on Build GRCh38
rs754172229 Chr.12: 75291860-75291860 on Build GRCh38
rs1057032116 Chr.12: 75291702-75291702 on Build GRCh38
rs762718498 Chr.12: 75291815-75291815 on Build GRCh38
rs749680359 Chr.12: 75291753-75291753 on Build GRCh38
rs143721078 Chr.12: 75291795-75291795 on Build GRCh38
rs757562779 Chr.12: 75291867-75291867 on Build GRCh38
rs149686440 Chr.12: 75291686-75291686 on Build GRCh38
rs755756973 Chr.12: 75291843-75291843 on Build GRCh38
rs1043491616 Chr.12: 75291747-75291747 on Build GRCh38
rs1012586089 Chr.12: 75291788-75291788 on Build GRCh38
rs375363191 Chr.12: 75291832-75291832 on Build GRCh38
rs780756820 Chr.12: 75291713-75291713 on Build GRCh38
rs769042371 Chr.12: 75291799-75291799 on Build GRCh38
rs778167450 Chr.12: 75291728-75291728 on Build GRCh38
rs766854192 Chr.12: 75291773-75291773 on Build GRCh38
rs752378232 Chr.12: 75291721-75291721 on Build GRCh38
rs754498841 Chr.12: 75291696-75291696 on Build GRCh38
rs200740298 Chr.12: 75291810-75291810 on Build GRCh38
rs201980880 Chr.12: 75291805-75291805 on Build GRCh38
rs768257695 Chr.12: 75291848-75291849 on Build GRCh38
rs766283289 Chr.12: 75291816-75291816 on Build GRCh38
rs895604079 Chr.12: 75291737-75291737 on Build GRCh38
rs747003889 Chr.12: 75291849-75291850 on Build GRCh38
rs771454252 Chr.12: 75291766-75291766 on Build GRCh38
rs369056810 Chr.12: 75291733-75291733 on Build GRCh38
rs759008413 Chr.12: 75291827-75291827 on Build GRCh38

View Full Product Details