Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 54650330-54650563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK5
Entrez Gene ID
55816
Gene Name
docking protein 5
Gene Aliases
C20orf180, IRS-6, IRS6
UniGene
Hs.473133
Gene Chromosome Location
Chr.20: 54475472-54651171 on Build GRCh38
Transcripts
NM_018431.4 NM_177959.2 XM_011528904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2763836 c.899C>T p.P300L Chr.20: 54650457-54650457 on Build GRCh38
3693589 c.899C>A p.P300Q Chr.20: 54650457-54650457 on Build GRCh38
3911624 c.873G>A p.L291L Chr.20: 54650431-54650431 on Build GRCh38
6160491 c.869C>T p.P290L Chr.20: 54650427-54650427 on Build GRCh38
7365302 c.871C>G p.L291V Chr.20: 54650429-54650429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779986265 Chr.20: 54650480-54650480 on Build GRCh38
rs148201917 Chr.20: 54650441-54650441 on Build GRCh38
rs778767434 Chr.20: 54650368-54650368 on Build GRCh38
rs956520491 Chr.20: 54650364-54650364 on Build GRCh38
rs747856502 Chr.20: 54650509-54650509 on Build GRCh38
rs373557448 Chr.20: 54650515-54650515 on Build GRCh38
rs371376809 Chr.20: 54650460-54650460 on Build GRCh38
rs567023193 Chr.20: 54650435-54650435 on Build GRCh38
rs201718507 Chr.20: 54650436-54650436 on Build GRCh38
rs771750294 Chr.20: 54650448-54650449 on Build GRCh38
rs375799502 Chr.20: 54650421-54650421 on Build GRCh38
rs746871266 Chr.20: 54650490-54650490 on Build GRCh38
rs929743069 Chr.20: 54650386-54650386 on Build GRCh38
rs776192777 Chr.20: 54650506-54650506 on Build GRCh38
rs745734024 Chr.20: 54650475-54650475 on Build GRCh38
rs763393546 Chr.20: 54650528-54650528 on Build GRCh38
rs761267752 Chr.20: 54650408-54650408 on Build GRCh38
rs776203787 Chr.20: 54650403-54650403 on Build GRCh38
rs79858797 Chr.20: 54650390-54650390 on Build GRCh38
rs1046853462 Chr.20: 54650388-54650388 on Build GRCh38
rs765750227 Chr.20: 54650416-54650416 on Build GRCh38
rs369209022 Chr.20: 54650496-54650496 on Build GRCh38
rs949258204 Chr.20: 54650397-54650397 on Build GRCh38
rs538040340 Chr.20: 54650507-54650507 on Build GRCh38
rs535580638 Chr.20: 54650505-54650505 on Build GRCh38
rs368274278 Chr.20: 54650380-54650380 on Build GRCh38
rs150282152 Chr.20: 54650464-54650464 on Build GRCh38
rs763478880 Chr.20: 54650425-54650425 on Build GRCh38
rs769142097 Chr.20: 54650512-54650512 on Build GRCh38
rs200965457 Chr.20: 54650468-54650468 on Build GRCh38
rs768254732 Chr.20: 54650399-54650399 on Build GRCh38
rs752802483 Chr.20: 54650461-54650461 on Build GRCh38
rs775430243 Chr.20: 54650381-54650381 on Build GRCh38
rs142008881 Chr.20: 54650442-54650442 on Build GRCh38

View Full Product Details