Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77941129-77941400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS4
Entrez Gene ID
92105
Gene Name
integrator complex subunit 4
Gene Aliases
INT4, MST093
UniGene
Hs.533723
Gene Chromosome Location
Chr.11: 77874491-77994849 on Build GRCh38
Transcripts
NM_033547.3 XM_011545352.2 XM_011545353.2 XM_017018560.1

SNP Detail

SNP ID Chromosome Location
rs771520611 Chr.11: 77941193-77941193 on Build GRCh38
rs778854118 Chr.11: 77941173-77941173 on Build GRCh38
rs552515694 Chr.11: 77941373-77941373 on Build GRCh38
rs770562087 Chr.11: 77941271-77941271 on Build GRCh38
rs573382092 Chr.11: 77941290-77941290 on Build GRCh38
rs769045816 Chr.11: 77941286-77941286 on Build GRCh38
rs775772945 Chr.11: 77941273-77941273 on Build GRCh38
rs564941166 Chr.11: 77941377-77941377 on Build GRCh38
rs575737587 Chr.11: 77941353-77941353 on Build GRCh38
rs777241872 Chr.11: 77941236-77941236 on Build GRCh38
rs776474958 Chr.11: 77941317-77941318 on Build GRCh38
rs746704736 Chr.11: 77941255-77941255 on Build GRCh38
rs949884971 Chr.11: 77941361-77941361 on Build GRCh38
rs112337042 Chr.11: 77941374-77941374 on Build GRCh38
rs762008610 Chr.11: 77941296-77941296 on Build GRCh38
rs909204182 Chr.11: 77941240-77941240 on Build GRCh38
rs763316544 Chr.11: 77941284-77941284 on Build GRCh38
rs542250703 Chr.11: 77941318-77941318 on Build GRCh38
rs545240253 Chr.11: 77941376-77941376 on Build GRCh38
rs747641435 Chr.11: 77941181-77941181 on Build GRCh38
rs767758530 Chr.11: 77941303-77941303 on Build GRCh38

View Full Product Details