Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17771519-17771762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC1
Entrez Gene ID
3746
Gene Name
potassium voltage-gated channel subfamily C member 1
Gene Aliases
EPM7, KV3.1, KV4, NGK2
UniGene
Hs.552896
Gene Chromosome Location
Chr.11: 17734810-17783055 on Build GRCh38
Transcripts
NM_001112741.1 NM_004976.4 XM_011520080.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4694198 c.601C>A p.L201M Chr.11: 17771695-17771695 on Build GRCh38
3979461 c.655G>C p.V219L Chr.11: 17771749-17771749 on Build GRCh38
6206652 c.652A>G p.I218V Chr.11: 17771746-17771746 on Build GRCh38
2092221 c.665C>T p.T222M Chr.11: 17771759-17771759 on Build GRCh38
7152071 c.617C>A p.T206N Chr.11: 17771711-17771711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752386683 Chr.11: 17771676-17771676 on Build GRCh38
rs371344625 Chr.11: 17771688-17771688 on Build GRCh38
rs887180585 Chr.11: 17771641-17771641 on Build GRCh38
rs1003601980 Chr.11: 17771564-17771564 on Build GRCh38
rs555448656 Chr.11: 17771555-17771555 on Build GRCh38
rs761762491 Chr.11: 17771579-17771579 on Build GRCh38
rs754387189 Chr.11: 17771659-17771659 on Build GRCh38
rs751102079 Chr.11: 17771653-17771653 on Build GRCh38
rs536785934 Chr.11: 17771544-17771544 on Build GRCh38
rs948267059 Chr.11: 17771543-17771543 on Build GRCh38
rs766186382 Chr.11: 17771630-17771630 on Build GRCh38
rs745968113 Chr.11: 17771730-17771730 on Build GRCh38
rs1005589935 Chr.11: 17771651-17771651 on Build GRCh38
rs79039048 Chr.11: 17771556-17771556 on Build GRCh38
rs940978237 Chr.11: 17771633-17771633 on Build GRCh38
rs761737637 Chr.11: 17771645-17771646 on Build GRCh38
rs374846297 Chr.11: 17771699-17771699 on Build GRCh38
rs772042572 Chr.11: 17771734-17771734 on Build GRCh38
rs762912737 Chr.11: 17771623-17771623 on Build GRCh38
rs779234164 Chr.11: 17771711-17771711 on Build GRCh38
rs991304915 Chr.11: 17771562-17771562 on Build GRCh38
rs757808689 Chr.11: 17771700-17771700 on Build GRCh38
rs573610082 Chr.11: 17771578-17771578 on Build GRCh38
rs907870726 Chr.11: 17771606-17771606 on Build GRCh38
rs767280558 Chr.11: 17771670-17771670 on Build GRCh38
rs552970887 Chr.11: 17771735-17771735 on Build GRCh38

View Full Product Details