Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27164958-27165215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA10-HOXA9
Entrez Gene ID
100534589
Gene Name
HOXA10-HOXA9 readthrough
Gene Aliases
-
UniGene
Hs.659350
Gene Chromosome Location
Chr.7: 27162438-27180261 on Build GRCh38
Transcripts
NR_037940.1 NM_152739.3

Gene Symbol
HOXA9
Entrez Gene ID
3205
Gene Name
homeobox A9
Gene Aliases
ABD-B, HOX1, HOX1.7, HOX1G
UniGene
Hs.659350
Gene Chromosome Location
Chr.7: 27162438-27165530 on Build GRCh38
Transcripts
NR_037940.1 NM_152739.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4751613 c.304G>C p.A102P Chr.7: 27165154-27165154 on Build GRCh38
6029285 c.243G>A p.A81A Chr.7: 27165215-27165215 on Build GRCh38
3769999 c.280C>A p.P94T Chr.7: 27165178-27165178 on Build GRCh38
3637742 c.382C>T p.R128W Chr.7: 27165076-27165076 on Build GRCh38
4690973 c.283delC p.Q95fs*60 Chr.7: 27165175-27165175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778792261 Chr.7: 27165193-27165193 on Build GRCh38
rs754905237 Chr.7: 27165143-27165143 on Build GRCh38
rs779145878 Chr.7: 27165085-27165085 on Build GRCh38
rs769367273 Chr.7: 27165158-27165158 on Build GRCh38
rs765931391 Chr.7: 27165043-27165043 on Build GRCh38
rs573758059 Chr.7: 27165161-27165161 on Build GRCh38
rs150821305 Chr.7: 27165002-27165002 on Build GRCh38
rs773091352 Chr.7: 27165030-27165030 on Build GRCh38
rs935619986 Chr.7: 27165020-27165020 on Build GRCh38
rs1040126912 Chr.7: 27165190-27165190 on Build GRCh38
rs567037909 Chr.7: 27165004-27165004 on Build GRCh38
rs766866768 Chr.7: 27165179-27165179 on Build GRCh38
rs1057051769 Chr.7: 27165132-27165132 on Build GRCh38
rs780585301 Chr.7: 27165136-27165136 on Build GRCh38
rs984989441 Chr.7: 27165152-27165152 on Build GRCh38
rs769817684 Chr.7: 27164988-27164988 on Build GRCh38
rs781712176 Chr.7: 27165051-27165051 on Build GRCh38
rs777951245 Chr.7: 27165152-27165154 on Build GRCh38
rs200450890 Chr.7: 27165154-27165154 on Build GRCh38
rs1000950303 Chr.7: 27165162-27165162 on Build GRCh38
rs147261225 Chr.7: 27165034-27165034 on Build GRCh38
rs749442438 Chr.7: 27165003-27165003 on Build GRCh38
rs762677209 Chr.7: 27165122-27165122 on Build GRCh38
rs746824728 Chr.7: 27165149-27165149 on Build GRCh38
rs768842404 Chr.7: 27165073-27165073 on Build GRCh38
rs754400549 Chr.7: 27165186-27165186 on Build GRCh38
rs143934224 Chr.7: 27165011-27165011 on Build GRCh38
rs374547527 Chr.7: 27165050-27165050 on Build GRCh38
rs199550076 Chr.7: 27164987-27164987 on Build GRCh38
rs200486389 Chr.7: 27165130-27165130 on Build GRCh38
rs748173136 Chr.7: 27165147-27165148 on Build GRCh38
rs767458451 Chr.7: 27165021-27165021 on Build GRCh38
rs368446662 Chr.7: 27165098-27165098 on Build GRCh38
rs750729932 Chr.7: 27165167-27165167 on Build GRCh38
rs967454305 Chr.7: 27165087-27165087 on Build GRCh38
rs767704574 Chr.7: 27165125-27165125 on Build GRCh38
rs761066551 Chr.7: 27165170-27165170 on Build GRCh38
rs750827678 Chr.7: 27165127-27165127 on Build GRCh38
rs750288349 Chr.7: 27164989-27164989 on Build GRCh38
rs752362815 Chr.7: 27165116-27165116 on Build GRCh38
rs748261114 Chr.7: 27165145-27165145 on Build GRCh38
rs771025386 Chr.7: 27165112-27165112 on Build GRCh38
rs777393633 Chr.7: 27165148-27165148 on Build GRCh38
rs138117599 Chr.7: 27165068-27165068 on Build GRCh38
rs763826633 Chr.7: 27165166-27165166 on Build GRCh38
rs745705983 Chr.7: 27165190-27165201 on Build GRCh38
rs559119879 Chr.7: 27165093-27165093 on Build GRCh38
rs569313107 Chr.7: 27165045-27165045 on Build GRCh38
rs761562140 Chr.7: 27165012-27165012 on Build GRCh38
rs201482064 Chr.7: 27165044-27165044 on Build GRCh38
rs760465140 Chr.7: 27165107-27165107 on Build GRCh38
rs759116325 Chr.7: 27165157-27165157 on Build GRCh38
rs368757601 Chr.7: 27165091-27165091 on Build GRCh38
rs772398323 Chr.7: 27165148-27165149 on Build GRCh38
rs755117477 Chr.7: 27165137-27165137 on Build GRCh38
rs562472814 Chr.7: 27165006-27165006 on Build GRCh38
rs755489506 Chr.7: 27165192-27165192 on Build GRCh38
rs770972572 Chr.7: 27165151-27165151 on Build GRCh38
rs35355140 Chr.7: 27165113-27165113 on Build GRCh38

View Full Product Details